    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • ANKRD42 Antibodies
          Invitrogen
          ANKRD42 Polyclonal Antibody
          View all (3) ANKRD42 antibodies
          Tech Support
          Cite ANKRD42 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          ANKRD42 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ANKRD42 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          ANKRD42 Antibody (PA5-69122) in WB
          Western blot analysis of ANKRD42 on NCI-H226 cells. The sample was probed with a ANKRD42 polyclonal antibody (Product # PA5-69122) using a primary antibody dilution of 0.2-1.0 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          ANKRD42 Antibody in Western Blot (WB)
          ANKRD42 Polyclonal Antibody
          Product Details
          PA5-69122
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          0.2-1.0 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          synthetic peptide directed towards the N terminal region of human ANKRD42
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          50 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS with 2% sucrose
          Contains
          0.09% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2689041
          Product Specific Information
          This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 93%
          Target Information
          ANKRD42 is a protein coding gene.
          Bioinformatics
          Protein Aliases: Ankyrin repeat domain-containing protein 42; protein phosphatase 1, regulatory subunit 79; SARP2; several ankyrin repeat protein; unnamed protein product
          View more View less
          Gene Aliases: ANKRD42; PPP1R79; SARP
          View more View less
          UniProt ID: (Human) Q8N9B4
          View more View less
          Entrez Gene ID: (Human) 338699
          View more View less
          Function(s)
          cyclin-dependent protein serine/threonine kinase inhibitor activity NF-kappaB binding
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0