Search Thermo Fisher Scientific
FIGURE: 1 / 4
Immunogen sequence: MKILLVFDFD NTIIDDNSDT WIVQCAPNKK LPIELRDSYR KGFWTEFMGR VFKYLGDKGV REHEMKRAVT SLPFTPGMVE LFNFIRKNKD KFDCIIISDS; Positive Samples: SH-SY5Y, A-549, H460
Phosphatase that has high activity toward pyridoxal 5'-phosphate (PLP). Also active at much lower level toward pyrophosphate, phosphoethanolamine (PEA), phosphocholine (PCho), phospho-l-tyrosine, fructose-6-phosphate, p-nitrophenyl phosphate, and h-glycerophosphate.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Protein Aliases: Pyridoxal phosphate phosphatase PHOSPHO2
Gene Aliases: 1700048E23Rik; AI661517; AU021728; AV006103; Phos2; PHOSPHO2
UniProt ID: (Human) Q8TCD6, (Mouse) Q9D9M5, (Rat) Q66HC4
Entrez Gene ID: (Human) 493911, (Mouse) 73373, (Rat) 295663
Molecular Function: hydrolase phosphatase metabolite interconversion enzyme