Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAAAAAAAA AGAAGGRGSG PGRRRHLVPG AGGEAGEGAP GGAGDYGNGL ESEELEPEEL LLEPEPEPEP EEEPPRPRAP PGAPGPGPGS GAPGSQEEEE
This gene encodes an abundant nuclear protein that binds with high affinity to nascent poly(A) tails. The protein is required for progressive and efficient polymerization of poly(A) tails at the 3' ends of eukaryotic transcripts and controls the size of the poly(A) tail to about 250 nt. At steady-state, this protein is localized in the nucleus whereas a different poly(A) binding protein is localized in the cytoplasm. This gene contains a GCG trinucleotide repeat at the 5' end of the coding region, and expansion of this repeat from the normal 6 copies to 8-13 copies leads to autosomal dominant oculopharyngeal muscular dystrophy (OPMD) disease. Related pseudogenes have been identified on chromosomes 19 and X. Read-through transcription also exists between this gene and the neighboring upstream BCL2-like 2 (BCL2L2) gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nuclear poly(A)-binding protein 1; PABII; PABP-2; poly(A) binding protein 2; poly(A) binding protein II; poly(A) binding protein, nuclear 1; poly(A)-binding protein 2; Poly(A)-binding protein II; Polyadenylate-binding nuclear protein 1; Polyadenylate-binding protein 2
Gene Aliases: mPABII; OPMD; PAB2; PABII; PABP-2; PABP2; Pabp3; PABPN1
UniProt ID: (Human) Q86U42, (Mouse) Q8CCS6
Entrez Gene ID: (Human) 8106, (Rat) 116697, (Mouse) 54196
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support