Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Pan SHANK Antibodies

          Rockland

          PAN-SHANK Monoclonal Antibody (N23b/49)

          View all (12) Pan SHANK antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PAN-SHANK Monoclonal Antibody (N23b/49)

          • Antibody Testing Data (1)
          PAN-SHANK Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PAN-SHANK Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PAN-SHANK Antibody (200-301-G42) in WB

          Western Blot of mouse anti-SHANK antibody. Lane 1: Rat Brain Membrane lysate. Primary antibody: SHANK antibody at 1:1,000 for overnight at 4°C. Secondary antibody: Goat anti-mouse IgG HRP secondary antibody at 1:10,000 for 45 min at RT. Block: 5% Blotto overnight 4°C. Predicted/Observed size: 158.6 kDa/160 kDa. Other band(s): none. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PAN-SHANK Antibody in Western Blot (WB)

          Product Details

          200-301-G42

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-10 µg/mL
          -

          Immunohistochemistry (IHC)

          0.1-1 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          1-10 µg/mL
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          N23b/49

          Immunogen

          SHANK Antibody was produced in mice by repeated immunizations raised against a fusion protein corresponding to an internal region of rat Shank2.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein G

          Storage buffer

          PBS, pH 7.2, with 50% glycerol

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          Shank proteins make up a family of scaffold proteins identified through their interaction with a variety of membrane and cytoplasmic proteins. Shank proteins at postsynaptic sites of excitatory synapses play roles in signal transmission into the postsynaptic neuron. Studies suggest that Shank2 is expressed in the neurons of the developing retina, and could play a role in the neuronal differentiation of the developing retina. Other recent studies suggest that the disruption of glutamate receptors at the Shank postsynaptic platform could contribute to the destruction of the postsynaptic density, which underlies the synaptic dysfunction and loss in Alzheimer's disease.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: cortactin SH3 domain-binding protein; Cortactin-binding protein 1; CortBP1; GKAP/SAPAP interacting protein; GKAP/SAPAP-interacting protein; proline rich synapse associated protein 1; proline rich synapse associated protein 2; proline-rich synapse associated protein 1; Proline-rich synapse-associated protein 1; Proline-rich synapse-associated protein 2; ProSAP1; ProSAP2; SAPAP-interacting protein synamon; SH3 and multiple ankyrin repeat domains 1; SH3 and multiple ankyrin repeat domains protein 1; SH3 and multiple ankyrin repeat domains protein 2; SH3 and multiple ankyrin repeat domains protein 3; SH3 and multiple ankyrin repeat domains protein 3 variant c-3; SH3 and multiple ankyrin repeat domains protein 3 variant c-4; SH3 and multiple ankyrin repeat domains-like protein 3 variant 1; SH3 and multiple ankyrin repeat domains-like protein 3 variant 2; SH3 and multiple ankyrin repeat domains-like protein 3 variant 3; SH3 and multiple ankyrin repeat domains-like protein 3 variant c-1; SH3 and multiple ankyrin repeat domains-like protein 3 variant c-2; SH3 and multiple ankyrin repeat domains-like protein 3 variant d-1; SH3 and multiple ankyrin repeat domains-like protein 3 variant d-2; SH3 and multiple ankyrin repeat domains-like protein 3 variant d-3; SH3 and multiple ankyrin repeat domains-like protein 3 variant e-1; SH3 and multiple ankyrin repeat domains-like protein 3 variant e-2; SH3/ankyrin domain gene 1; SH3/ankyrin domain gene 2; SH3/ankyrin domain gene 3; shank postsynaptic density protein; Shank postsynaptic density protein 3a; Shank1; Shank2; Shank3; shank3 splice variant II; shank3 splice variant III; shank3 splice variant IV; shank3e splice variant III; shank3e splice variant IV; Somatostatin receptor-interacting protein; SPANK-1; SPANK-2; SPANK-3; SSTR-interacting protein; Synamon

          View more View less

          Gene Aliases: AI841104; AUTS17; CORTBP1; CTTNBP1; DEL22q13.3; KIAA1022; Kiaa1650; mKIAA1022; PROSAP1; PROSAP2; PSAP2; SCZD15; SHANK; SHANK1; Shank1a; SHANK2; Shank3; Shank3b; SHANK3c-3; SHANK3c-4; SPANK-1; SPANK-2; SPANK-3; Spank1; SSTRIP; synamon

          View more View less

          UniProt ID: (Human) Q9Y566, (Human) Q9UPX8, (Rat) Q9WV48, (Mouse) D3YZU1, (Mouse) Q80Z38, (Rat) Q9QX74, (Mouse) Q4ACU6, (Rat) Q9JLU4

          View more View less

          Entrez Gene ID: (Human) 50944, (Human) 22941, (Human) 85358, (Rat) 78957, (Mouse) 243961, (Mouse) 210274, (Rat) 171093, (Mouse) 58234, (Rat) 59312

          View more View less

          Function(s)
          protein binding protein C-terminus binding SH3 domain binding GKAP/Homer scaffold activity somatostatin receptor binding protein complex binding ionotropic glutamate receptor binding identical protein binding ankyrin repeat binding scaffold protein binding actin binding zinc ion binding receptor signaling complex scaffold activity protein self-association
          Process(es)
          protein complex assembly cytoskeletal anchoring at plasma membrane long-term memory associative learning adult behavior negative regulation of actin filament bundle assembly social behavior protein localization to synapse olfactory behavior habituation neuromuscular process controlling balance determination of affect righting reflex synapse maturation dendritic spine morphogenesis positive regulation of dendritic spine development vocalization behavior regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity positive regulation of excitatory postsynaptic potential MAPK cascade embryonic epithelial tube formation synapse assembly learning memory striatal medium spiny neuron differentiation locomotory exploration behavior vocal learning negative regulation of cell volume positive regulation of long-term neuronal synaptic plasticity brain morphogenesis protein oligomerization positive regulation of synapse structural plasticity positive regulation of synaptic transmission, glutamatergic regulation of dendritic spine morphogenesis postsynaptic density assembly AMPA glutamate receptor clustering NMDA glutamate receptor clustering guanylate kinase-associated protein clustering regulation of long-term synaptic potentiation positive regulation of long-term synaptic potentiation positive regulation of glutamate receptor signaling pathway regulation of long term synaptic depression regulation of grooming behavior regulation of behavioral fear response positive regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity exploration behavior long-term synaptic potentiation long term synaptic depression nervous system development cell differentiation brain development response to nutrient
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-48z6k:80/100.66.78.162:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0