Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
PAN3 is functions in cytoplasmic mRNA decay. It is as part of the Pan nuclease complex, recruits polyadenylate-binding protein which in turn stimulates PAN2 nuclease activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PAB-dependent poly(A)-specific ribonuclease subunit 3; PAB1P-dependent poly(A)-nuclease; PAB1P-dependent poly(A)-specific ribonuclease; PABP-dependent poly(A) nuclease 3; PABP1-dependent poly A-specific ribonuclease subunit PAN3; PAN deadenylation complex subunit 3; PAN2-PAN3 deadenylation complex subunit PAN3; PAN3 poly(A) specific ribonuclease subunit homolog; Poly(A)-nuclease deadenylation complex subunit 3; RP11-179F17.3
Gene Aliases: PAN3
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support