Applications | Tested Dilution | Publications |
---|---|---|
Immunohistochemistry (Paraffin) (IHC (P)) |
1:50-200 | - |
Immunohistochemistry (Frozen) (IHC (F)) |
1:50-200 | - |
Immunocytochemistry (ICC/IF) |
1:50-200 | - |
Product Specifications | |
---|---|
Species Reactivity |
Mouse, Rat |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
KLH conjugated synthetic peptide derived from human PAX6 |
Conjugate |
Alexa Fluor™ 488 |
Excitation/Emission Profile |
View spectra |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Protein A |
Storage buffer |
TBS with 50% glycerol, 1% BSA |
Contains |
0.03% Proclin 300 |
Storage conditions |
-20°C or -80°C if preferred |
This gene encodes paired box gene 6, one of many human homologs of the Drosophila melanogaster gene prd. In addition to the hallmark feature of this gene family, a conserved paired box domain, the encoded protein also contains a homeo box domain. Both domains are known to bind DNA, and function as regulators of gene transcription. This gene is expressed in the developing nervous system, and in developing eyes. Mutations in this gene are known to cause ocular disorders such as aniridia and Peter's anomaly. Alternatively spliced transcript variants encoding either the same or different isoform have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aniridia; Dickie's small eye; Keratitis; MGC17209; Oculorhombin; paired box gene 6; Paired Box Gene-6; paired box homeotic gene 6; Paired box protein Pax-6; small eye
Gene Aliases: 1500038E17Rik; AEY11; Dey; Gsfaey11; Pax-6; Pax6; Sey
UniProt ID: (Rat) P63016, (Mouse) P63015
Entrez Gene ID: (Rat) 25509, (Mouse) 18508
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support