Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:500-1:1,000 | - |
ELISA (ELISA) |
1 ng/mL | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Mouse / IgG2a, kappa |
Class |
Monoclonal |
Type |
Antibody |
Clone |
3H7 |
Immunogen |
PAX8 (AAH01060, 1 a.a. approximately 450 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
See Label |
Purification |
Affinity chromatography |
Storage buffer |
PBS, pH 7.4 |
Contains |
no preservative |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
Sequence of this protein is as follows: MPHNSIRSGH GGLNQLGGAF VNGRPLPEVV RQRIVDLAHQ GVRPCDISRQ LRVSHGCVSK ILGRYYETGS IRPGVIGGSK PKVATPKVVE KIGDYKRQNP TMFAWEIRDR LLAEGVCDND TVPSVSSINR IIRTKVQQPF NLPMDSCVAT KSLSPGHTLI PSSAVTPPES PQSDSLGSTY SINGLLGIAQ PGSDKRKMDD SDQDSCRLSI DSQSSSSGPR KHLRTDAFSQ HHLEPLECPF ERQHYPEAYA SPSHTKGEQG LYPLPLLNST LDDGKATLTP SNTPLGRNLS THQTYPVVAD PHSPFAIKQE TPEVSSSSST PSSLSSSAFL DLQQVGSGVP PFNAFPHAAS VYGQFTGQAL LSGREMVGPT LPGYPPHIPT SGQGSYASSA IAGMVAGSEY SGNAYGHTPY SSYSEAWRFP NSSLLSSPYY YSSTSRPSAP PTTATAFDHL
This gene encodes a member of the paired box family of transcription factors. Members of this gene family typically encode proteins that contain a paired box domain, an octapeptide, and a paired-type homeodomain. This nuclear protein is involved in thyroid follicular cell development and expression of thyroid-specific genes. Mutations in this gene have been associated with thyroid dysgenesis, thyroid follicular carcinomas and atypical follicular thyroid adenomas. Alternatively spliced transcript variants encoding different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HGNC:8622; Paired box protein Pax-8; paired domain gene 8
Gene Aliases: PAX8
UniProt ID: (Human) Q06710
Entrez Gene ID: (Human) 7849
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support