The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections.
Involved in transcriptional activation and repression of select genes by chromatin remodeling (alteration of DNA-nucleosome topology).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2310032M22Rik; BAF180; BRG1-associated factor 180; hPB1; Polybromo-1D; Protein polybromo-1
Gene Aliases: 2610016F04Rik; AI507524; BAF180; PB1; PBRM1
UniProt ID: (Human) Q86U86
Entrez Gene ID: (Human) 55193, (Mouse) 66923
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support