Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 2
Immunogen sequence: KMEEYDDVNL LTADFQLLFN NAKSYYKPDS PEYKAACKLW DLYLRTRNEF VQKGEADDED DDEDGQDNQG TVTEGSSPAY LKEILEQLLE AIVVATNPSG RLISELFQKL PSKVQYPDYY AIIKEPIDLK TIAQRIQNGS YKSIHA
Involved in transcriptional activation and repression of select genes by chromatin remodeling (alteration of DNA-nucleosome topology).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BAF180; BRG1-associated factor 180; hPB1; Polybromo-1D; Protein polybromo-1
Gene Aliases: BAF180; PB1; PBRM1
UniProt ID: (Human) Q86U86
Entrez Gene ID: (Human) 55193
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support