Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:1,000 | View 1 publication 1 publication |
Immunoprecipitation (IP) |
1:100 | - |
ChIP assay (ChIP) |
1:25 | - |
Product Specifications | |
---|---|
Species Reactivity |
Bovine, Horse, Human, Mouse, Non-human primate, Rat |
Published species |
Human |
Host/Isotype |
Rabbit / IgG |
Class |
Monoclonal |
Type |
Antibody |
Clone |
E.107.0 |
Immunogen |
Synthetic peptide corresponding to the amino terminus of human PCAF protein |
Conjugate |
Unconjugated |
Form |
Liquid |
Purification |
Affinity chromatography |
Storage buffer |
0.01M HEPES, pH 7.5, with 0.15M NaCl, 100µg/mL BSA, 50% glycerol |
Contains |
<0.02% sodium azide |
Storage conditions |
-20°C |
RRID |
AB_10978155 |
It is not recommended to aliquot this antibody.
This antibody is not cross-reactive with the related GCN5L2 protein.
PCAF (histone acetyltransferase KAT2B) functions as a histone acetyltransferase (HAT) to promote transcriptional activation. It has significant histone acetyltransferase activity with core histones (H3 and H4) and nucleosome core particles. PCAF also acetylates non-histone proteins such as ACLY, PLK4, and TBX5. It inhibits cell-cycle progression and counteracts the mitogenic activity of the adenoviral oncoprotein E1A. PCAF also acts as a circadian transcriptional coactivator which enhances the activity of the circadian transcriptional activators NPAS2-ARNTL/BMAL1 and CLOCK-ARNTL/BMAL1 heterodimers. It is involved in heart and limb development by mediating acetylation of TBX5 and acetylation regulating nucleocytoplasmic shutting of TBX5.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CREBBP-associated factor; Histone acetylase PCAF; Histone acetyltransferase KAT2B; Histone acetyltransferase PCAF; K(lysine) acetyltransferase 2B; Lysine acetyltransferase 2B; P/ PCAF; P/CAF; P300/CBP-associated factor; Spermidine acetyltransferase KAT2B
Gene Aliases: A930006P13Rik; AI461839; AW536563; CAF; KAT2B; P/CAF; PCAF
UniProt ID: (Human) Q92831, (Mouse) Q9JHD1
Entrez Gene ID: (Horse) 100061976, (Bovine) 407215, (Human) 8850, (Rat) 301164, (Mouse) 18519
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support