Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
This monoclonal antibody CSTEM31 reacts with human Protocadherin-1, also known as PCDH1 and cadherin-like protein 1.
Applications Reported: This CSTEM31 antibody has been reported for use in immunocytochemical staining and flow cytometric analysis. (Fluorochrome-conjugated CSTEM31 is recommended for use in flow cytometery.)
Applications Tested: This CSTEM31 antibody has been tested by immunocytochemistry on formaldehyde-fixed and permeabilized human iPSCs, and by flow cytometry on human iPSCs. For immunocytochemistry, this can be used at less than or equal to 5 µg/mL. For flow cytometry, this antibody can be used at less than or equal to 0.5 µg/test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Protocadherin 1, also known as PCDH1 and cadherin-like protein 1, is a transmembrane protein involved in cell-cell adhesion. Protocadherin 1 belongs to a group of cadherin-like proteins termed delta1-protocadherins, which consists of protocadherin-1, -7, -9, and -11(X/Y). The delta1-protocadherins have seven extracellular cadherin-repeats, a transmembrane domain, and a cytoplasmic domain, and can be expressed in two isoforms, short or long. Expression of protocadherin 1 is regulated both temporally and spatially, and plays a role both in early development and mature tissues. In mice, it is expressed in a number of tissues and organs, including the brain, spinal cord, peripheral ganglia, inner ear, hair follicles, kidney, reproductive organs, prostate, and the seminal gland. Notably, protocadherin 1 is a susceptibility gene for Bronchial Hyperresponsiveness in humans. Expression of protocadherin 1 has also been identified on human pluripotent stem cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cadherin-like 1; Cadherin-like protein 1; PC42; protocadherin 42; Protocadherin-1; Protocadherin-42
Gene Aliases: PC42; PCDH1; PCDH42
UniProt ID: (Human) Q08174
Entrez Gene ID: (Human) 5097
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support