Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:2,000-1:20,000 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Mouse / IgG1 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
2C11B8 |
Immunogen |
PDGFRA Fusion Protein Ag1385 (1-209 aa encoded by BC015186) |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
0.85 mg/mL |
Purification |
Caprylic acid precipitation |
Storage buffer |
PBS, pH 7.3, with 50% glycerol |
Contains |
0.1% sodium azide |
Storage conditions |
-20°C |
Immunogen sequence: MGTSHPAFL VLGCLLTGLS LILCQLSLPS ILPNENEKVV QLNSSFSLRC FGESEVSWQY PMSEEESSDV EIRNEENNSG LFVTVLEVSS ASAAHTGLYT CYYNHTQTEE NELEGRHIYI YVPDPDVAFV PLGMTDYLVI VEDDDSAIIP CRTTDPETPV TLHNSEGVVP ASYDSRQGFN GTFTVGPYIC EATVKGKKFQ TIPFNVYALK (1-209 aa encoded by BC015186)
PDGFRA is a cell surface tyrosine kinase receptor for members of the platelet-derived growth factor family. These growth factors are mitogens for cells of mesenchymal origin. The identity of the growth factor bound to a receptor monomer determines whether the functional receptor is a homodimer or a heterodimer, composed of both platelet-derived growth factor receptor alpha and beta polypeptides. Studies in knockout mice, where homozygosity is lethal, indicate that the alpha form of the platelet-derived growth factor receptor is particularly important for kidney development since mice heterozygous for the receptor exhibit defective kidney phenotypes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha platelet-derived growth factor receptor; Alpha-type platelet-derived growth factor receptor; CD140 antigen-like family member A; CD140a; CD140a antigen; MGC74795; OTTHUMP00000218656; PDGF A-chain; PDGF Receptor alpha; PDGF-R-alpha; PDGFR-2; pdgfr-a; PDGFRA/BCR fusion; Platelet-derived growth factor alpha receptor; Platelet-derived growth factor receptor 2; Platelet-derived growth factor receptor alpha; platelet-derived growth factor receptor, alpha polypeptide; rearranged-in-hypereosinophilia-platelet derived growth factor receptor alpha fusion protein
Gene Aliases: CD140A; GAS9; PDGFR-2; PDGFR2; PDGFRA; RHEPDGFRA
UniProt ID: (Human) Q9UD28
Entrez Gene ID: (Human) 5156
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support