Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DDSFSEAHS EFLKAASNLR DNYRFAHTNV ESLVNEYDDN GEGIILFRPS HLTNKFEDKT VAYTEQKMTS GKIKKFIQEN IFGICPHMTE DNKDLIQGKD LLIAYYDVDY EKNAKGSNYW RNRVMMVAKK FLDAGHKLNF AVASRKTFSH ELSDFGLEST AGEIPVVAIR TAKGEKFVMQ EEFSRDGKAL ERFLQDYFDG NLKRYLKSEP IPESNDGPVK VVVAENFDEI VNNENKDVLI EFYAPWCGHC KNLEPKYKEL GEKLSKDPNI VIAKMDATAN DVPSPYEVRG FPTIYFSPAN KKLNPKKYEG GRELSDFISY LQREATNPPV IQEEKPKKKK KAQEDL (161-505 aa encoded by BC014433)
This gene encodes a protein of the endoplasmic reticulum that interacts with lectin chaperones calreticulin and calnexin to modulate folding of newly synthesized glycoproteins. The protein was once thought to be a phospholipase; however, it has been demonstrated that the protein actually has protein disulfide isomerase activity. It is thought that complexes of lectins and this protein mediate protein folding by promoting formation of disulfide bonds in their glycoprotein substrates.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 58 kDa glucose-regulated protein; 58 kDa microsomal protein; Disulfide isomerase ER-60; endoplasmic reticulum P58; endoplasmic reticulum protein; Endoplasmic reticulum resident protein 57; Endoplasmic reticulum resident protein 60; epididymis secretory protein Li 269; epididymis secretory sperm binding protein Li 93n; ER p57; ER protein 57; ER protein 60; ER-60 protease; ERp 57; ERp60; glucose regulated protein, 58 kDa; glucose regulated protein, 58kDa; GRP 57; GRP 58; HIP-70; oxidoreductase ERp57; p58; PDIA 3; phospholipase C, alpha; phospholipase C-alpha; PI PLC; protein disulfide isomerase associated 3; protein disulfide isomerase family A, member 3; protein disulfide isomerase-associated 3; Protein disulfide-isomerase A3; Q-2
Gene Aliases: 58kDa; ER60; Erp; ERP57; ERP60; ERp61; GRP57; GRP58; HEL-S-269; HEL-S-93n; HsT17083; P58; PDI; PDI-Q2; PDIA3; PI-PLC; PLC[a]; Plca
UniProt ID: (Human) Q8IYF8, (Mouse) P27773, (Rat) P11598
Entrez Gene ID: (Human) 2923, (Mouse) 14827, (Rat) 29468
Molecular Function: chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support