Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Control: Mouse spleen tissue, rat cerebellum tissue, human tonsil tissue, human lung cancer tissue, human kidney tissue, mouse testis tissue, A549, SH-SY-5Y.
Presenilins, which are components of the gamma-secretase protein complex, are required for intramembranous processing of some type I transmembrane proteins, such as the Notch proteins and the beta-amyloid precursor protein. Signaling by Notch receptors mediates a wide range of developmental cell fates. Processing of the beta-amyloid precursor protein generates neurotoxic amyloid beta peptides, the major component of senile plaques associated with Alzheimer's disease. This gene encodes a protein that is required for Notch pathway signaling, and for the activity and accumulation of gamma-secretase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gamma-secretase subunit PEN-2; hematopoietic stem/progenitor cells protein MDS033; presenilin enhancer 2 homolog; presenilin enhancer gamma secretase subunit; Presenilin enhancer protein 2
Gene Aliases: MDS033; MSTP064; PEN-2; PEN2; PSENEN
UniProt ID: (Human) Q9NZ42
Entrez Gene ID: (Human) 55851
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support