Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
The MW of this protein is 15 kDa, and this antibody specifically recognizes the 15 kDa protein.
Immunogen sequence: MAGWQSYVD NLMCDGCCQE AAIVGYCDAK YVWAATAGGV FQSITPIEID MIVGKDREGF FTNGLALGAK KCSVIRDSLY VDGDCTMDIR TKSQGGEPTY NVAVGRAGRV LVFVMGKEGV HGGGLNKKAY SMAKYLRDSG F (1-140 aa encoded by BC018049)
Profilin 2 is a ubiquitous actin monomer-binding protein belonging to the profilin family. It is thought to regulate actin polymerization in response to extracellular signals.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actin binding protein; profilin II; profilin-2-like; unnamed protein product
Gene Aliases: D3S1319E; PFL; Pfn
UniProt ID: (Human) P35080, (Mouse) Q9JJV2, (Rat) Q9EPC6
Entrez Gene ID: (Human) 5217, (Pig) 100621523, (Mouse) 18645, (Rat) 81531
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support