Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • PGAM1 Antibodies

          Proteintech

          PGAM1 Monoclonal Antibody (2H2A9), CoraLite® Plus 594

          View all (17) PGAM1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite PGAM1 Monoclonal Antibody (2H2A9), CoraLite® Plus 594

          • Antibody Testing Data (1)
          PGAM1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PGAM1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PGAM1 Antibody (CL594-67470) in ICC/IF

          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using CoraLite® Plus 594 PGAM1 antibody (CL594-67470, Clone: 2H2A9) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PGAM1 Antibody in Immunocytochemistry (ICC/IF)

          Product Details

          CL594-67470

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          2H2A9

          Immunogen

          PGAM1 fusion protein Ag9250

          Conjugate

          CoraLite® Plus 594 CoraLite® Plus 594 CoraLite® Plus 594
          • Unconjugated
          • Request custom conjugation

          Excitation/Emission Max

          588/604 nm

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein G

          Storage buffer

          PBS, pH 7.3, with 50% glycerol, 0.5% BSA

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, store in dark

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: MAAYKLVLI RHGESAWNLE NRFSGWYDAD LSPAGHEEAK RGGQALRDAG YEFDICFTSV QKRAIRTLWT VLDAIDQMWL PVVRTWRLNE RHYGGLTGLN KAETAAKHGE AQVKIWRRSY DVPPPPMEPD HPFYSNISKD RRYADLTEDQ LPSCESLKDT IARALPFWNE EIVPQIKEGK RVLIAAHGNS LRGIVKHLEG LSEEAIMELN LPTGIPIVYE LDKNLKPIKP MQFLGDEETV RKAMEAVAAQ GKAKK

          Target Information

          PGAM1 belongs to the phosphoglycerate mutase family. PGAM1 is an important component of glucose and 2,3-BPGA (2,3-bisphosphoglycerate) metabolism and catalyzes the reversible reaction of 3-phosphoglycerate (3-PGA) to 2-phosphoglycerate (2-PGA) in the glycolytic pathway. The PGAM1 protein is a dimeric enzyme containing, in different tissues, different proportions of a slow-migrating muscle (MM) isozyme, a fast-migrating brain (BB) isozyme, and a hybrid form (MB). Mutations in PGAM1 cause muscle phosphoglycerate mutase efficiency, also known as glycogen storage disease X. Recombinant human PGAM protein, fused to His-tag at N-terminus, was expressed in E. coli and purified by using conventional chromatography techniques.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: BPG-dependent PGAM 1; epididymis secretory protein Li 35; PGAM-B; Phosphoglycerate mutase 1; phosphoglycerate mutase 1 (brain); phosphoglycerate mutase A, nonmuscle form; Phosphoglycerate mutase isozyme B; RP11-452K12.8

          View more View less

          Gene Aliases: 2310050F24Rik; CDABP0006; HEL-S-35; Pgam-1; PGAM-B; PGAM1; PGAMA; Pgmut

          View more View less

          UniProt ID: (Human) P18669, (Rat) P25113, (Mouse) Q9DBJ1

          View more View less

          Entrez Gene ID: (Human) 5223, (Rat) 24642, (Mouse) 18648

          View more View less

          Function(s)
          bisphosphoglycerate mutase activity bisphosphoglycerate 2-phosphatase activity phosphoglycerate mutase activity protein binding protein kinase binding 2,3-bisphosphoglycerate-dependent phosphoglycerate mutase activity catalytic activity hydrolase activity isomerase activity intramolecular transferase activity, phosphotransferases mutase
          Process(es)
          gluconeogenesis glycolytic process regulation of glycolytic process dephosphorylation regulation of pentose-phosphate shunt respiratory burst canonical glycolysis metabolic process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-56756dfb8b-mgmkt:80/100.66.75.22:80.
          git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.1-2025.06.25-1.0