Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MVATEDERLA GSGEGERLDF LRDRHVRFFQ RCLQVLPERY SSLETSRLTI AFFALSGLDM LDSLDVVNKD DIIEWIYSLQ VLPTEDRSNL NRCGFRGSSY LGIPF
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CTC-428G20.3; Geranylgeranyl transferase type I subunit beta; Geranylgeranyl transferase type-1 subunit beta; geranylgeranyltransferase type 1 beta; geranylgeranyltransferase type I (GGTase-I); geranylgeranyltransferase type I beta subunit; GGTase-I-beta; protein geranylgeranyltransferase type I, beta subunit; Type I protein geranyl-geranyltransferase subunit beta
Gene Aliases: BGGI; GGTI; PGGT1B
UniProt ID: (Human) P53609, (Rat) P53610
Entrez Gene ID: (Human) 5229, (Rat) 81746
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support