Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PIN1 Antibodies

          Abnova

          PIN1 Recombinant Rabbit Monoclonal Antibody

          View all (38) PIN1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PIN1 Recombinant Rabbit Monoclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          PIN1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          PIN1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PIN1 Antibody (RAB01998) in IHC (P)

          Immunohistochemical staining (Formalin/PFA-fixed paraffin-embedded sections) of human breast cancer with PIN1 recombinant monoclonal antibody, clone R01-7G8 (Product # RAB01998). High-pressure and temperature Sodium Citrate pH 6.0 was used for antigen retrieval. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PIN1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PIN1 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          RAB01998

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:100
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          Immunoprecipitation (IP)

          1:20
          -
          Product Specifications

          Species Reactivity

          Hamster, Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          CHO cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Immunogen

          Recombinant protein corresponding to human PIN1.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Purification

          Affinity chromatography

          Storage buffer

          50mM tris glycine, pH 7.4, with 40% glycerol, 150mM NaCl, 0.05% BSA

          Contains

          0.01% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          PIN1 (Peptidylprolyl Cis/Trans Isomerase, NIMA-Interacting 1) is a Protein Coding gene. Diseases associated with PIN1 include Meibomian Cyst and Viral Exanthem. Among its related pathways are Transport of the SLBP independent Mature mRNA and Cytokine Signaling in Immune system. Gene Ontology (GO) annotations related to this gene include peptidyl-prolyl cis-trans isomerase activity and mitogen-activated protein kinase binding.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: EC 5.2.1.8; NIMA-interacting; parvulin PIN1; peptidyl-prolyl cis-trans isomerase Pin1; peptidylprolyl cis/trans isomerase, NIMA-interacting 1; PPIase Pin1; prolyl isomerase; protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1; protein interacting with never in mitosis A1; rotamase Pin1; UBL5; unnamed protein product

          View more View less

          Gene Aliases: 0610025L01Rik; D9Bwg1161e; DOD; UBL5

          View more View less

          UniProt ID: (Human) Q13526, (Mouse) Q9QUR7

          View more View less

          Entrez Gene ID: (Human) 5300, (Rat) 298696, (Mouse) 23988, (Chinese hamster) 100773691

          View more View less

          Function(s)
          peptidyl-prolyl cis-trans isomerase activity motor activity protein binding beta-catenin binding cis-trans isomerase activity mitogen-activated protein kinase kinase binding GTPase activating protein binding tau protein binding phosphoserine binding phosphothreonine binding phosphoprotein binding ubiquitin ligase activator activity isomerase activity chaperone
          Process(es)
          protein peptidyl-prolyl isomerization response to hypoxia positive regulation of protein phosphorylation regulation of mitotic nuclear division Rho protein signal transduction regulation of gene expression neuron differentiation negative regulation of transforming growth factor beta receptor signaling pathway regulation of protein stability protein destabilization regulation of cytokinesis negative regulation of protein catabolic process positive regulation of transcription from RNA polymerase II promoter microtubule polymerization synapse organization protein stabilization negative regulation of SMAD protein import into nucleus negative regulation of ERK1 and ERK2 cascade protein localization to mitochondrion cellular response to hypoxia positive regulation of canonical Wnt signaling pathway regulation of protein localization to nucleus negative regulation of beta-amyloid formation negative regulation of brown fat cell differentiation negative regulation of cell motility regulation of cell proliferation negative regulation of neuron apoptotic process positive regulation of neuron apoptotic process positive regulation of cell growth involved in cardiac muscle cell development regulation protein catabolic process at postsynapse
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch