Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MRPDDKYHEK QAQVEATKLK QKVEALSPGD RQQIYEKGLE LRSQQSKPQD ASCLPALKVS DIEPTIPVTE LDVVLTAGDI PVQYCAQPTN GMVYFRAFSS LNTLPEELRP YVPLFCSVLT KLGCGLLDYR EQAQQIELKT GGMSASPHVL PDDSHMDTYE QGVLFSSLCL DRNLPDMMQL WSEIFNNPCF EEEEHFKVLV KMTAQELANG IPDSGHLYAS IRAGRTLTPA GDLQETFSGM DQVRLMKRIA EMTDIKPILR KLPRIKKHLL NGDNMRCSVN ATPQQMPQTE KAVEDFLRSI GRSKKERRPV RPHTVEKPVP SSSGGDAHVP HGSQVIRKLV MEPTFKPWQM KTHFLMPFPV NYVGECIRTV PYTDPDHASL KILARLMTAK FLHTEIREKG GAYGGGAKLS HNGIFTLYSY RDPNTIETLQ SFGKAVDWAK SGKFTQQDID EAKLSVFSTV DAPVAPSDKG MDHFLYGLSD EMKQAHREQL FAVSHDKLLA VSDRYLGTGK STHGLAILGP ENPKIAKDPS WIIR
The protein encoded by this gene is an ATP-dependent metalloprotease that degrades post-cleavage mitochondrial transit peptides. The encoded protein binds zinc and can also degrade amyloid beta A4 protein, suggesting a possible role in Alzheimer's disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPreP; metalloendoprotease; Metalloprotease 1; metalloprotease 1 (pitrilysin family); Pitrilysin metalloproteinase 1; PreP peptidasome; Presequence protease, mitochondrial; unnamed protein product
Gene Aliases: KIAA1104; MP1; PITRM1; PREP; SCAR30
UniProt ID: (Human) Q5JRX3
Entrez Gene ID: (Human) 10531
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support