Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
PLEKHG2 may be a transforming oncogene with exchange activity for CDC42. PLEKHG2 may be a guanine-nucleotide exchange factor (GEF) for RAC1 and CDC42.PLEKHG2 activated by the binding to subunits beta and gamma of the heterotrimeric guanine nucleotide-binding protein (G protein).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: common-site lymphoma/leukemia guanine nucleotide exchange factor; CTB-60E11.4; DKFZp667J2325; FLJ00018; FLJ22458; FLJ38638; PH domain-containing family G member 2; pleckstrin homology domain containing, family G (with RhoGef domain) member 2; Pleckstrin homology domain-containing family G member 2
Gene Aliases: ARHGEF42; CLG; LDAMD; PLEKHG2
UniProt ID: (Human) Q9H7P9
Entrez Gene ID: (Human) 64857
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support