Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Immunogen sequence: SSPAKNFFKK AAAALFGGKK DKARYIDTHN RVSKEDEDIY KLRHDLKKTS ITQQPSKHRT DEELQPPTTT VARSGTPAVE NKQQIGDAIR MIVRGTLGSC SSSSECLEDS TMGSVADTVA RVLRGCLENM PEADCIPKEQ L
PLK2 is a serum inducible serine/threonine kinase that is a member of the polo family of serine/threonine kinases which have a role in normal cell division. The kinase may play a role in embryogenesis, wound healing, and neoplasia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: h PLK-2; hSNK; PLK-2; Polo-like kinase 2; Serine/threonine-protein kinase PLK2; Serine/threonine-protein kinase SNK; Serum-inducible kinase
Gene Aliases: hPlk2; hSNK; PLK2; SNK
UniProt ID: (Human) Q9NYY3
Entrez Gene ID: (Human) 10769
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support