Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence:
MNARGLGSE LKDSIPVTEL SASGPFESHD LLRKGFSCVK NELLPSHPLE LSEKNFQLNQ DKMNFSTLRN IQGLFAPLKL QMEFKAVQQV QRLPFLSSSN LSLDVLRGND ETIGFEDILN DPSQSEVMGE PHLMVEYKLG LL
Molecular chaperone essential for the assembly of standard proteasomes and immunoproteasomes. Degraded after completion of proteasome maturation. Mediates the association of 20S preproteasome with the endoplasmic reticulum.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2510048O06Rik; hUMP1; Proteasome maturation protein; Proteassemblin; Protein UMP1 homolog; Voltage-gated K channel beta subunit 4.1; voltage-gated potassium channel beta subunit 4.1
Gene Aliases: C13orf12; HSPC014; HSPC036; PNAS-110; POMP; RGD1305831; UMP1
UniProt ID: (Human) Q9Y244
Entrez Gene ID: (Human) 51371, (Rat) 288455
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support