Applications | Tested Dilution | Publications |
---|---|---|
Immunoprecipitation (IP) |
8-15 µL/mg lysate | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Between 113 and 163 |
Conjugate |
Unconjugated |
Form |
liquid |
Concentration |
1 mg/mL |
Storage conditions |
4° C |
The recommended shelf life for this product is 1 year from date of receipt.
Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5'-ends (PubMed:11413139, PubMed:30454648). Also a component of the MRP ribonuclease complex, which cleaves pre-rRNA sequences (PubMed:28115465). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPop5; processing of precursor 5, ribonuclease P/MRP subunit; Ribonuclease P/MRP protein subunit POP5
Gene Aliases: AD-008; HSPC004; POP5; x0003
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support