Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:500-1:2,000 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Rat |
Host/Isotype |
Rabbit / IgG |
Class |
Recombinant Monoclonal |
Type |
Antibody |
Clone |
6 |
Immunogen |
Recombinant Mouse PPM1A/PP2CA Protein |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Protein A |
Storage buffer |
PBS |
Contains |
no preservative |
Storage conditions |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
RRID |
AB_2785573 |
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
This antibody has specificity for Mouse PPM1A/PP2CA.
PPM1A (Protein phosphatase 1A) is also named as PPPM1A and belongs to the PP2C family. It is a serine/threonine protein phosphatase that is essential for regulating cellular stress responses in eukaryotes. PPM1A plays an important role in tumor suppression and that it negatively controls many signaling pathways which are critical for trophoblast invasion. This protein distributes in both the cytoplasm and nucleus of mammalian cells. It has 2 isoforms produced by alternative splicing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IA; Mpp alpha; PP2C-alpha; Protein phosphatase 1A; protein phosphatase 1A (formerly 2C), magnesium-dependent, alpha isoform; protein phosphatase 1A, magnesium dependent, alpha isoform; Protein phosphatase 2C isoform alpha; Protein phosphatase IA; Protein phosphatase type 1A (formely 2C) Mg-dependent alpha isoform; Protein phosphatase type 1A (formely 2C), Mg-dependent, alpha isoform; protein phosphatase, Mg2+/Mn2+ dependent, 1A
Gene Aliases: 2310003C21Rik; 2900017D14Rik; AI427932; AU017636; MMPa-2; MPPa-1; PP2C-ALPHA; Pp2c1; PP2CA; PP2Calpha; PPM1A; PPPM1A
UniProt ID: (Human) P35813, (Mouse) P49443, (Rat) P20650
Entrez Gene ID: (Human) 5494, (Mouse) 19042, (Rat) 24666
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support