Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PRDM9 Antibodies

          Invitrogen

          PRDM9 Recombinant Rabbit Monoclonal Antibody (1H9Z1)

          View all (10) PRDM9 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PRDM9 Recombinant Rabbit Monoclonal Antibody (1H9Z1)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          PRDM9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          PRDM9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PRDM9 Antibody (MA5-51196) in IHC (P)

          Immunohistochemistry analysis of PRDM9 in paraffin-embedded human colon carcinoma. Samples were incubated with PRDM9 Recombinant monoclonal antibody (Product # MA5-51196) using a dilution of 1:800 (40x lens). Perform high pressure antigen retrieval with 10 mM citrate buffer pH 6.0 before commencing with IHC staining protocol. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PRDM9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PRDM9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PRDM9 Recombinant Rabbit Monoclonal Antibody (1H9Z1)

          Product Details

          MA5-51196

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:500-1:1,000
          -

          ELISA (ELISA)

          1 µg/mL
          -

          CUT&RUN (CUT&RUN)

          105 cells /1 µg
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          1H9Z1

          Immunogen

          Recombinant protein (or fragment)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 0.05% BSA, 50% glycerol

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_3094253

          Product Specific Information

          Immunogen sequence: DSEDSDEEWT PKQQVSPPWV PFRVKHSKQQ KESSRMPFSG ESNVKEGSGI ENLLNTSGSE HVQKPVSSLE EGNTSGQHSG KKLKLRKKNV EVKMYRLRER KGLAYEEVSE PQDDDYLYCE KCQNFFIDSC PNHGPPLFVK DSMVDRGHPN HSVLSLPPGL RISPSGIPEA GLGVWNEASD LPVGLHFGPY EGQITEDEEA ANSGYSWLIT KGRNCYEYVD GQDESQANWM RYVNCARDDE EQNLVAFQYH RKIFYRTCRV IRPGCELLVW YGDEYGQELG IKWG

          Target Information

          The PR domain is a protein-protein interaction module of about 100 amino acids. PR domain-containing proteins, such as PRDM9, are often involved in transcriptional regulation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: [histone H3]-lysine36 N-trimethyltransferase PRDM9; [histone H3]-lysine4 N-trimethyltransferase PRDM9; [histone H3]-lysine9 N-trimethyltransferase PRDM9; [histone H4]-lysine20 N-methyltransferase PRDM9; [histone H4]-N-methyl-L-lysine20 N-methyltransferase PRDM9; histone methyl transferase; Histone-lysine N-methyltransferase PRDM9; minisatellite binding protein 3 (115kD); PFM6; PR domain 9; PR domain containing 9; PR domain zinc finger protein 9; PR domain-containing protein 9; Protein-lysine N-methyltransferase PRDM9; unnamed protein product

          View more View less

          Gene Aliases: KMT8B; MEISETZ; MSBP3; PFM6; PRDM9; ZNF899

          View more View less

          UniProt ID: (Human) Q9NQV7

          View more View less

          Entrez Gene ID: (Human) 56979

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding DNA binding methyltransferase activity zinc ion binding recombination hotspot binding protein-lysine N-methyltransferase activity transferase activity histone methyltransferase activity histone methyltransferase activity (H3-K4 specific) protein homodimerization activity sequence-specific DNA binding metal ion binding histone methyltransferase activity (H3-K36 specific) histone H4K20me methyltransferase activity histone H4K20 monomethyltransferase activity histone H3K9 trimethyltransferase activity histone H3K36 trimethyltransferase activity histone H3K4 trimethyltransferase activity
          Process(es)
          meiotic gene conversion chromatin organization chromatin remodeling regulation of transcription, DNA-templated synapsis female gamete generation regulation of gene expression positive regulation of reciprocal meiotic recombination methylation reciprocal DNA recombination negative regulation of apoptotic process male gamete generation meiotic cell cycle meiosis I cell cycle process positive regulation of fertilization double-strand break repair involved in meiotic recombination
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6479c7f679-26whj:80/100.66.74.142:80.
          git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.50.0-Offline