Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:2,000 | - |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:50 | - |
Immunocytochemistry (ICC/IF) |
1:100 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Mouse / IgG1 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
OTI9A3 |
Immunogen |
Full length human recombinant protein of human PRDX4 produced in HEK293T cell. |
Conjugate |
Unconjugated |
Form |
lyophilized |
Concentration |
1 mg/mL |
Purification |
Affinity chromatography |
Storage buffer |
PBS, pH 7.3, with 8% trehalose |
Contains |
no preservative |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Peroxiredoxin (Prx) is a growing peroxidase family, whose mammalian members have been known to connect with cell proliferation, differentiation, and apoptosis. Many isoforms (about 50 proteins), collected in accordance to the amino acid sequence homology, containing active site cysteine residue, and the thiol-specific antioxidant activity, distribute throughout all the kingdoms. Among them, mammalian Prx consists of 6 different members grouped into typical 2-Cys, atypical 2-Cys Prx, and 1-Cys Prx. Except Prx VI belonging to 1-Cys Prx subgroup, the other five 2-Cys Prx isotypes have the thioredoxin-dependent peroxidase (TPx) activity utilizing thioredoxin, thioredoxin reductase, and NADPH as a reducing system. Mammalian Prxs are 20-30 kilodalton in molecular size and vary in subcellular localization: Prx I, II, and VI in cytosol, Prx III in mitochondria, Prx IV in ER and secretion, Prx V showing complicated distribution including peroxisome, mitochondria and cytosol.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Antioxidant enzyme AOE372; AOE37-2; EC 1.11.1.15; epididymis secretory sperm binding protein Li 97n; Peroxiredoxin IV; Peroxiredoxin-4; Prx 4; Prx-IV; Thioredoxin dependent peroxide reductase A0372; thioredoxin peroxidase (antioxidant enzyme); Thioredoxin peroxidase AO372; Thioredoxin-dependent peroxide reductase A0372; Thioredoxin-dependent peroxiredoxin 4
Gene Aliases: AOE37-2; AOE372; HEL-S-97n; PRDX4; PRX-4
UniProt ID: (Human) Q13162
Entrez Gene ID: (Human) 10549
Molecular Function:
peroxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support