Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1-2 µg/mL | - |
Immunocytochemistry (ICC/IF) |
Assay-dependent | - |
ELISA (ELISA) |
Assay-dependent | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Rat |
Host/Isotype |
Mouse / IgG3 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
PRMT3-367 |
Immunogen |
Synthetic peptide: TVHKNKKDPRSLIV conjugated to KLH, corresponding to amino acids 501-514 of Rat PRMT3 |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
2.1 mg/mL |
Purification |
Protein A |
Storage buffer |
0.01M PBS, pH 7.4 |
Contains |
15mM sodium azide |
Storage conditions |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
RRID |
AB_2171674 |
MA1-25469 is expected to cross react with mouse (100% conserved), rat (100% conserved), and human (92% conserved) due to sequence homology.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Type I protein arginine N-methyltransferases (PRMTs), such as PRMT3, catalyze the formation of asymmetric N(G),N(G)-dimethylarginine (ADMA) residues in proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: heterogeneous nuclear ribonucleoprotein methyltransferase-like 3; Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 3; HMT1 hnRNP methyltransferase-like 3; protein arginine methyltransferase 3; Protein arginine N-methyltransferase 3; protein arginine N-methyltransferase 3(hnRNP methyltransferase S. cerevisiae)-like 3
Gene Aliases: 2010005E20Rik; 2410018A17Rik; AL033309; HRMT1L3; PRMT3
UniProt ID: (Human) O60678, (Rat) O70467, (Mouse) Q922H1
Entrez Gene ID: (Human) 10196, (Rat) 89820, (Mouse) 71974
Molecular Function:
protein modifying enzyme
zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support