Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: VAFPISREAF QALEKLNNRQ LNYVQLEIDI KNEIIILANT TNTELKDLPK RIPKDSARYH FFLYKHSHEG DYLESIVFIY SMPGYTCSIR ERMLYSSCKS RLLEIVERQL QMDVIRKIEI DNGDELTADF LYEEVHPKQH AHKQSSAKPK GPAGKRGIRR LIRGPAETEA TTD
This gene encodes twinfilin, an actin monomer-binding protein conserved from yeast to mammals. Studies of the mouse counterpart suggest that this protein may be an actin monomer-binding protein, and its localization to cortical G-actin-rich structures may be regulated by the small GTPase RAC1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A6 protein tyrosine kinase; Protein A6; Protein tyrosine kinase 9; PTK9 protein tyrosine kinase 9; twinfilin actin-binding protein 1; twinfilin, actin-binding protein, homolog 1; Twinfilin-1
Gene Aliases: A6; PTK9; TWF1
UniProt ID: (Human) Q12792, (Rat) Q5RJR2
Entrez Gene ID: (Human) 5756, (Rat) 315265
Molecular Function: non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support