Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For maximum recovery of product, centrifuge the vial prior to removing the cap.
Pan-KChIP refers to the collective group of Kv Channel-Interacting Proteins (KChIPs), which are auxiliary subunits that associate with voltage-gated potassium (Kv) channels. These proteins modulate the trafficking, surface expression, and functional properties of Kv channels, thereby influencing neuronal excitability and signaling. Dysregulation of KChIPs has been implicated in various neurological conditions, including epilepsy and ataxia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A-type potassium channel modulating protein 3; A-type potassium channel modulatory protein 1; A-type potassium channel modulatory protein 1.2; A-type potassium channel modulatory protein 2; A-type potassium channel modulatory protein 3; A-type potassium channel modulatory protein 4; A-type potassium channel modulatory protein KCNIP1; A-type potassium channel modulatory protein KCNIP2; Calsenilin; calsenilin presenilin-binding protein EF hand transcription factor; calsenilin, presenilin binding protein, EF hand transcription factor; calsenilin, presenilin-binding protein, EF hand transcription fa; calsenilin, presenilin-binding protein, EF hand transcription factor; Calsenilin-like protein; Cardiac voltage-gated potassium channel modulatory subunit; downstream regulatory element-antagonist modulator; DRE-antagonist modulator; DREAM; KChIP1; KChIP2; KChIP3; KChIP4; Kv channel interacting protein 1; Kv channel interacting protein 2; Kv channel interacting protein 3; Kv channel interacting protein 3, calsenilin; Kv channel interacting protein 4; Kv channel-interacting protein 1; Kv channel-interacting protein 2; Kv channel-interacting protein 3; Kv channel-interacting protein 4; potassium channel auxiliary subunit KCHIP2; potassium channel interacting protein 1; potassium channel interacting protein 3; potassium channel interacting protein 4; Potassium channel-interacting protein 1; Potassium channel-interacting protein 2; Potassium channel-interacting protein 4; Vesicle APC-binding protein
Gene Aliases: 4933407H12Rik; AI413860; AV032399; CALP; Calp250; CSEN; DREAM; KCHIP1; Kchip1.2; KCHIP2; KCHIP3; KCHIP4; KChIP4a; KCNIP1; KCNIP2; KCNIP3; KCNIP4; R74849; rKChIP3; VABP
UniProt ID: (Human) Q9NZI2, (Human) Q9NS61, (Human) Q9Y2W7, (Human) Q6PIL6, (Mouse) Q9JJ57, (Rat) Q8R426, (Mouse) Q9JJ69, (Rat) Q9JM59, (Mouse) Q9QXT8, (Rat) Q9JM47, (Rat) Q99MG9, (Mouse) Q6PHZ8
Entrez Gene ID: (Human) 30820, (Human) 30819, (Human) 30818, (Human) 80333, (Mouse) 70357, (Rat) 65023, (Mouse) 80906, (Rat) 56817, (Mouse) 56461, (Rat) 65199, (Rat) 259243, (Mouse) 80334
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support