Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Pan-KChIP Antibodies

          Antibodies Inc

          Pan-KChIP K+ Channel Monoclonal Antibody (K55/82), FL594

          View all (5) Pan-KChIP antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Pan-KChIP K+ Channel Monoclonal Antibody (K55/82), FL594

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          Pan-KChIP K+ Channel Monoclonal Antibody (K55/82), FL594

          Product Details

          75-006-FL594

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          K55/82

          Immunogen

          Fusion protein amino acids 1-216 (full length) of rat KChIP1b (accession number Q8R426) produced recombinantly in E. Coli.
          3D Epitope / Immunogen

          Conjugate

          FL594 FL594 FL594
          • Unconjugated
          • FL490
          • FL550
          • FL650
          • Request custom conjugation

          Excitation/Emission Max

          594/615 nm

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Dry ice (international)

          Product Specific Information

          For maximum recovery of product, centrifuge the vial prior to removing the cap.

          Target Information

          Pan-KChIP refers to the collective group of Kv Channel-Interacting Proteins (KChIPs), which are auxiliary subunits that associate with voltage-gated potassium (Kv) channels. These proteins modulate the trafficking, surface expression, and functional properties of Kv channels, thereby influencing neuronal excitability and signaling. Dysregulation of KChIPs has been implicated in various neurological conditions, including epilepsy and ataxia.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: A-type potassium channel modulating protein 3; A-type potassium channel modulatory protein 1; A-type potassium channel modulatory protein 1.2; A-type potassium channel modulatory protein 2; A-type potassium channel modulatory protein 3; A-type potassium channel modulatory protein 4; A-type potassium channel modulatory protein KCNIP1; A-type potassium channel modulatory protein KCNIP2; alternatively spliced; calcium binding protein; Calsenilin; calsenilin presenilin-binding protein EF hand transcription factor; calsenilin, presenilin binding protein, EF hand transcription factor; calsenilin, presenilin-binding protein, EF hand transcription fa; calsenilin, presenilin-binding protein, EF hand transcription factor; Calsenilin-like protein; Cardiac voltage-gated potassium channel modulatory subunit; downstream regulatory element-antagonist modulator; DRE-antagonist modulator; DREAM; KChIP; KChIP1; KChIP2; KCHIP2.2; KChIP3; KChIP4; KChIP4bs; KCNIp1; KCNIP1-Ia#II; KCNIP1-Ib; KCNIP1-Ib#II; KCNIP1-Ib+75inV; KCNIP2-Ia; KCNIP2-Ia#IIab; KCNIP2-Ia#IIb; KCNIP2-Ib#IIab; KCNIP2-Ic#IIab; KCNIP3-Ia; KCNIP3-Ib#II; KCNIP4-Ia; KCNIP4-Ia#II; KCNIP4-Ib#II; KCNIP4-Ic#II; KCNIP4-Id#II; KCNIP4-Ie#II; Kv channel interacting protein 1; Kv channel interacting protein 2; Kv channel interacting protein 3, calsenilin; Kv channel interacting protein 4; Kv channel-interacting protein 1; Kv channel-interacting protein 2; Kv channel-interacting protein 3; Kv channel-interacting protein 4; neuronal calcium sensor-1 homolog; potassium channel auxiliary subunit KCHIP2; potassium channel interacting protein 1; potassium channel interacting protein 3; potassium channel interacting protein 4; potassium channel interacting protein; KChIP2; Potassium channel-interacting protein 1; Potassium channel-interacting protein 2; Potassium channel-interacting protein 4; recoverin-family member; unnamed protein product; VABP protein; Vesicle APC-binding protein

          View more View less

          Gene Aliases: 4933407H12Rik; AI413860; AV032399; CALP; Calp250; CSEN; DREAM; KCHIP1; Kchip1.2; KCHIP2; KCHIP3; KCHIP4; KChIP4a; KCNIP1; KCNIP2; KCNIP3; KCNIP4; R74849; rKChIP3; VABP

          View more View less

          UniProt ID: (Human) Q9NZI2, (Human) Q9NS61, (Human) Q9Y2W7, (Human) Q6PIL6, (Mouse) Q9JJ57, (Rat) Q8R426, (Mouse) Q9JJ69, (Rat) Q9JM59, (Mouse) Q9QXT8, (Rat) Q9JM47, (Rat) Q99MG9, (Mouse) Q6PHZ8

          View more View less

          Entrez Gene ID: (Human) 30820, (Human) 30819, (Human) 30818, (Human) 80333, (Mouse) 70357, (Rat) 65023, (Mouse) 80906, (Rat) 56817, (Mouse) 56461, (Rat) 65199, (Rat) 259243, (Mouse) 80334

          View more View less

          Function(s)
          potassium channel activity calcium ion binding protein binding potassium channel regulator activity ion channel binding metal ion binding A-type (transient outward) potassium channel activity identical protein binding macromolecular complex binding ER retention sequence binding RNA polymerase II core promoter proximal region sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding voltage-gated ion channel activity protein heterodimerization activity protein N-terminus binding transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding DNA binding protein C-terminus binding sequence-specific DNA binding calcium-dependent protein binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter transcription, DNA-templated regulation of transcription, DNA-templated transcription from RNA polymerase II promoter transport ion transport potassium ion transport intracellular protein transport apoptotic process sensory perception of pain regulation of ion transmembrane transport regulation of neuron apoptotic process negative regulation of transcription, DNA-templated response to pain behavioral response to pain potassium ion transmembrane transport protein localization to plasma membrane regulation of potassium ion transmembrane transport regulation of signal transduction ion transmembrane transport signal transduction action potential detection of calcium ion muscle contraction chemical synaptic transmission regulation of heart contraction clustering of voltage-gated potassium channels regulation of membrane repolarization membrane repolarization membrane repolarization during cardiac muscle cell action potential regulation of potassium ion export across plasma membrane positive regulation of potassium ion export across plasma membrane potassium ion export protein heterooligomerization positive regulation of action potential potassium ion export across plasma membrane biological_process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline