Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • KVbeta1/2/3 Antibodies

          Antibodies Inc

          Pan-Kvbeta K+ Channel Monoclonal Antibody (K40/17)

          View all (10) KVbeta1/2/3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Pan-Kvbeta K+ Channel Monoclonal Antibody (K40/17)

          Pan-Kvbeta K+ Channel Monoclonal Antibody (K40/17)

          Product Details

          75-106

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          -

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          K40/17

          Immunogen

          Fusion protein amino acids 1-401 (full-length) of rat Kv1 (accession number P63144) produced recombinantly in E. Coli.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4

          Contains

          0.065% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Dry ice (international)

          Product Specific Information

          For maximum recovery of product, centrifuge the vial prior to removing the cap.

          Target Information

          KVbeta1, KVbeta2, and KVbeta3 are beta subunits of voltage-gated potassium (KV) channels. These auxiliary subunits modulate the properties and functions of the KV channel's alpha subunits. These KVbeta subunits are essential for the fine-tuning of KV channel activity, impacting the electrical properties of cells and contributing to the regulation of physiological processes such as neuronal signaling and cardiac rhythm.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: hKvbeta2; K(+) channel subunit beta-1; K(+) channel subunit beta-2; K(+) channel subunit beta-3; K+ channel Beta1a chain; Kv-beta-1; Kv-beta-2; Kv-beta-3; Neuroimmune protein F5; potassium channel beta 3 chain; potassium channel beta3 subunit; potassium channel shaker chain beta 1a; potassium channel, voltage gated shaker-related subfamily A regulatory beta subunit 2; potassium channel, voltage gated shaker-related subfamily A regulatory beta subunit 3; potassium channel, voltage gated subfamily A regulatory beta subunit 1; potassium channel, voltage gated subfamily A regulatory beta subunit 2; potassium channel, voltage gated subfamily A regulatory beta subunit 3; potassium channel, voltage-dependent, beta-3 subunit; potassium voltage gated channel shaker related subfamily beta member 1; potassium voltage gated channel, shaker related subfamily, beta member 1; potassium voltage gated channel, shaker related subfamily, beta member 2; potassium voltage gated channel, shaker related subfamily, beta member 3; potassium voltage-gated channel, shaker-related subfamily, beta member 1; potassium voltage-gated channel, shaker-related subfamily, beta member 2; potassium voltage-gated channel, shaker-related subfamily, beta member 3; RCK beta3; voltage-gated potassium channel beta subunit; Voltage-gated potassium channel subunit beta-1; Voltage-gated potassium channel subunit beta-2; Voltage-gated potassium channel subunit beta-3

          View more View less

          Gene Aliases: AKR6A3; AKR6A5; AKR6A9; Akr8a8; C330022D06Rik; Ckbeta2; Ckbeta3; F5; hKvb3; HKvbeta2; HKvbeta2.1; HKvbeta2.2; hKvBeta3; I2rf5; KCNA1B; KCNA2B; KCNA3.1B; KCNA3B; KCNAB1; KCNAB2; KCNAB3; Kcnab4; Kcnb3; KCNK2; KV-BETA-1; KV-BETA-2; KV-BETA-3; Kvb1; Kvb1.3; Kvbeta1.1; KvBeta3; mKv(beta)1; mKv(beta)4

          View more View less

          UniProt ID: (Human) Q14722, (Human) Q13303, (Human) O43448, (Mouse) P63143, (Rat) P63144, (Mouse) P62482, (Rat) P62483, (Rat) Q63494

          View more View less

          Entrez Gene ID: (Human) 7881, (Human) 8514, (Human) 9196, (Mouse) 16497, (Rat) 29737, (Mouse) 16498, (Rat) 29738, (Rat) 58981, (Mouse) 16499

          View more View less

          Function(s)
          aldo-keto reductase (NADP) activity voltage-gated potassium channel activity potassium channel regulator activity ion channel binding NADPH binding C-3 sterol dehydrogenase (C-4 sterol decarboxylase) activity isocitrate dehydrogenase activity mevaldate reductase activity voltage-gated ion channel activity protein binding gluconate dehydrogenase activity steroid dehydrogenase activity oxidoreductase activity epoxide dehydrogenase activity 5-exo-hydroxycamphor dehydrogenase activity 2-hydroxytetrahydrofuran dehydrogenase activity acetoin dehydrogenase activity phenylcoumaran benzylic ether reductase activity D-xylose:NADP reductase activity L-arabinose:NADP reductase activity D-arabinitol dehydrogenase, D-ribulose forming (NADP+) activity steroid dehydrogenase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor steroid dehydrogenase activity, acting on the CH-CH group of donors (R)-(-)-1,2,3,4-tetrahydronaphthol dehydrogenase activity 3-hydroxymenthone dehydrogenase activity very long-chain-3-hydroxyacyl-CoA dehydrogenase activity dihydrotestosterone 17-beta-dehydrogenase activity (R)-2-hydroxyisocaproate dehydrogenase activity L-arabinose 1-dehydrogenase (NADP+) activity L-xylulose reductase (NAD+) activity 3-ketoglucose-reductase activity (R)-2-hydroxyglutarate dehydrogenase activity D-arabinitol dehydrogenase, D-xylulose forming (NADP+) activity
          Process(es)
          potassium ion transport regulation of ion transmembrane transport potassium ion transmembrane transport hematopoietic progenitor cell differentiation neuromuscular process oxidation-reduction process NADPH oxidation regulation of potassium ion transmembrane transport regulation of protein localization to cell surface transport ion transport protein heterooligomerization learning or memory transmembrane transport regulation of potassium ion transmembrane transporter activity regulation of delayed rectifier potassium channel activity negative regulation of delayed rectifier potassium channel activity brain development heart development skeletal muscle tissue development myoblast differentiation diaphragm development
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7988999796-smrv7:80/100.66.72.156:80.
          git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.39.1-2025.11.99-1.0