Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For maximum recovery of product, centrifuge the vial prior to removing the cap.
Pan-MAGUK refers to the collective group of Membrane-Associated Guanylate Kinase (MAGUK) family proteins, which are scaffolding proteins involved in organizing signaling complexes at cellular membranes. These proteins play critical roles in synaptic signaling, cell adhesion, and maintaining the structure of tight junctions. Dysregulation of MAGUK proteins has been linked to various neurological disorders and cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Channel-associated protein of synapse-110; channel-associated protein of synapses, 110kDa; Chapsyn-110; discs large 2; discs large homolog 1; discs large homolog 1, scribble cell polarity complex component; discs large homolog 2; discs large homolog 3; discs large homolog 4; discs, large homolog 1; discs, large homolog 1 (Drosophila) transcript variant 6-v1; discs, large homolog 1 (Drosophila) transcript variant 6-v2; discs, large homolog 2; discs, large homolog 2, chapsyn-110; discs, large homolog 3; discs, large homolog 4; Disks large homolog 1; Disks large homolog 2; Disks large homolog 3; Disks large homolog 4; Drosophila discs-large tumor suppressor homologue (synapse associated protein); E-dlg/SAP97; Embryo-dlg/synapse-associated protein 97; hDlg; membrane associated guanylate kinase; MAGUK; Neuroendocrine-DLG; post-synaptic density protein 95; Postsynaptic density protein 93; Postsynaptic density protein 95; Postsynaptic density protein PSD-93; presynaptic protein SAP97; protein phosphatase 1, regulatory subunit 58; protein phosphatase 1, regulatory subunit 82; PSD-93; PSD-95; PSD-95 alpha 2b; PSD-95 beta; PSD-95/SAP90-related protein 1; SAP-102; SAP-90; SAP-97; SAP-97A; SAP102; SAP97A; Synapse-associated protein 102; Synapse-associated protein 90; Synapse-associated protein 97; Synapse-associated protein 97A; synapse-associated protein SAP90; synaptic density protein PSD-93; Tax interaction protein 15; XLMR
Gene Aliases: A330103J02Rik; B130052P05Rik; B230218P12Rik; B330007M19Rik; chapsyn-110; dJ1061C18.1.1; DLG1; DLG2; DLG3; DLG4; DLGH1; Dlgh2; Dlgh3; Dlgh4; E-dlg/SAP97; fa66c08; fa66e02; fd02c12; fu95c12; Gm1197; hdlg; im:7138640; KIAA1232; mKIAA1232; mKIAA4187; MRX; MRX90; NEDLG; PPP1R58; PPP1R82; PSD-93; PSD-95; PSD93; PSD95; SAP-90; SAP-97; SAP-97A; SAP102; SAP90; SAP90A; SAP97; si:ch211-276g21.1; wu:fa66c08; wu:fa66e02; wu:fc34d02; wu:fd02c12; wu:fu95c12; XLMR; zgc:55810
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support