Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Pan-MAGUK Antibodies

          Antibodies Inc

          Pan-MAGUK Monoclonal Antibody (K28/86), FL594

          View all (5) Pan-MAGUK antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Pan-MAGUK Monoclonal Antibody (K28/86), FL594

          Product Details

          75-029-FL594

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat, Xenopus, Zebrafish

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          K28/86

          Immunogen

          Fusion protein amino acids 77-299 of human PSD-95/SAP90 produced recombinantly in E. Coli.
          View immunogen

          Conjugate

          FL594 FL594 FL594
          • Unconjugated
          • FL490
          • FL550
          • FL650
          • Request custom conjugation

          Excitation/Emission Max

          594/615 nm

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Dry ice (international)

          Product Specific Information

          For maximum recovery of product, centrifuge the vial prior to removing the cap.

          Target Information

          Pan-MAGUK refers to the collective group of Membrane-Associated Guanylate Kinase (MAGUK) family proteins, which are scaffolding proteins involved in organizing signaling complexes at cellular membranes. These proteins play critical roles in synaptic signaling, cell adhesion, and maintaining the structure of tight junctions. Dysregulation of MAGUK proteins has been linked to various neurological disorders and cancers.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Channel-associated protein of synapse-110; channel-associated protein of synapses, 110kDa; Chapsyn-110; discs large 2; discs large homolog 1; discs large homolog 1, scribble cell polarity complex component; discs large homolog 2; discs large homolog 3; discs large homolog 4; discs, large homolog 1; discs, large homolog 1 (Drosophila) transcript variant 6-v1; discs, large homolog 1 (Drosophila) transcript variant 6-v2; discs, large homolog 2; discs, large homolog 2, chapsyn-110; discs, large homolog 3; discs, large homolog 4; Disks large homolog 1; Disks large homolog 2; Disks large homolog 3; Disks large homolog 4; Drosophila discs-large tumor suppressor homologue (synapse associated protein); E-dlg/SAP97; Embryo-dlg/synapse-associated protein 97; hDlg; membrane associated guanylate kinase; MAGUK; Neuroendocrine-DLG; post-synaptic density protein 95; Postsynaptic density protein 93; Postsynaptic density protein 95; Postsynaptic density protein PSD-93; presynaptic protein SAP97; protein phosphatase 1, regulatory subunit 58; protein phosphatase 1, regulatory subunit 82; PSD-93; PSD-95; PSD-95 alpha 2b; PSD-95 beta; PSD-95/SAP90-related protein 1; SAP-102; SAP-90; SAP-97; SAP-97A; SAP102; SAP97A; Synapse-associated protein 102; Synapse-associated protein 90; Synapse-associated protein 97; Synapse-associated protein 97A; synapse-associated protein SAP90; synaptic density protein PSD-93; Tax interaction protein 15; XLMR

          View more View less

          Gene Aliases: A330103J02Rik; B130052P05Rik; B230218P12Rik; B330007M19Rik; chapsyn-110; dJ1061C18.1.1; DLG1; DLG2; DLG3; DLG4; DLGH1; Dlgh2; Dlgh3; Dlgh4; E-dlg/SAP97; fa66c08; fa66e02; fd02c12; fu95c12; Gm1197; hdlg; im:7138640; KIAA1232; mKIAA1232; mKIAA4187; MRX; MRX90; NEDLG; PPP1R58; PPP1R82; PSD-93; PSD-95; PSD93; PSD95; SAP-90; SAP-97; SAP-97A; SAP102; SAP90; SAP90A; SAP97; si:ch211-276g21.1; wu:fa66c08; wu:fa66e02; wu:fc34d02; wu:fd02c12; wu:fu95c12; XLMR; zgc:55810

          View more View less

          UniProt ID: (Human) Q12959, (Human) Q15700, (Human) Q92796, (Human) P78352, (Rat) Q62696, (Mouse) Q811D0, (Mouse) Q91XM9, (Rat) Q63622, (Rat) Q62936, (Mouse) P70175, (Rat) P31016, (Mouse) Q62108, (Zebrafish) Q5PYH6, (Zebrafish) Q5PYH7

          View more View less

          Entrez Gene ID: (Human) 1739, (Human) 1740, (Human) 1741, (Human) 1742, (Rat) 25252, (Mouse) 13383, (Mouse) 23859, (Rat) 64053, (Rat) 58948, (Mouse) 53310, (Rat) 29495, (Mouse) 13385, (Zebrafish) 114446, (Zebrafish) 497638, (Zebrafish) 564081, (Western clawed frog) 779559, (Zebrafish) 100000823

          View more View less

          Function(s)
          guanylate kinase activity phosphoprotein phosphatase activity protein binding protein C-terminus binding cytoskeletal protein binding ligand-gated ion channel activity potassium channel regulator activity protein kinase binding phosphatase binding mitogen-activated protein kinase kinase binding protein complex scaffold ionotropic glutamate receptor binding ion channel binding L27 domain binding cadherin binding involved in cell-cell adhesion kinase binding protein phosphatase binding PDZ domain binding ubiquitin protein ligase binding Ras guanyl-nucleotide exchange factor activity beta-1 adrenergic receptor binding D1 dopamine receptor binding P2Y1 nucleotide receptor binding protein complex binding acetylcholine receptor binding neuroligin family protein binding scaffold protein binding protein heterodimerization activity protein domain specific binding receptor binding glutamate receptor binding structural molecule activity neurexin family protein binding scaffold/adaptor protein
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter branching involved in ureteric bud morphogenesis immunological synapse formation endothelial cell proliferation lens development in camera-type eye T cell cytokine production protein dephosphorylation actin filament organization mitotic cell cycle checkpoint establishment or maintenance of cell polarity chemical synaptic transmission nervous system development positive regulation of cell proliferation regulation of cell shape viral process single organismal cell-cell adhesion peristalsis positive regulation of actin filament polymerization cortical actin cytoskeleton organization astral microtubule organization membrane raft organization regulation of myelination activation of protein kinase activity ion transmembrane transport cellular protein complex localization T cell activation negative regulation of T cell proliferation regulation of membrane potential amyloid precursor protein metabolic process receptor clustering positive regulation of potassium ion transport cortical microtubule organization establishment or maintenance of epithelial cell apical/basal polarity negative regulation of mitotic cell cycle GMP metabolic process GDP metabolic process reproductive structure development embryonic skeletal system morphogenesis smooth muscle tissue development negative regulation of epithelial cell proliferation establishment of centrosome localization negative regulation of protein kinase B signaling hard palate development negative regulation of ERK1 and ERK2 cascade bicellular tight junction assembly protein localization to plasma membrane positive regulation of establishment of protein localization to plasma membrane receptor localization to synapse regulation of NIK/NF-kappaB signaling regulation of sodium ion transmembrane transport negative regulation of p38MAPK cascade negative regulation of phosphatase activity sensory perception of pain establishment of planar polarity negative regulation of cell proliferation MAPK cascade negative regulation of receptor internalization protein complex assembly signal transduction positive regulation of cytosolic calcium ion concentration learning synaptic vesicle maturation social behavior protein localization to synapse locomotory exploration behavior positive regulation of GTPase activity establishment of protein localization regulation of long-term neuronal synaptic plasticity positive regulation of synaptic transmission neuromuscular process controlling balance dendritic spine morphogenesis vocalization behavior AMPA glutamate receptor clustering regulation of N-methyl-D-aspartate selective glutamate receptor activity positive regulation of excitatory postsynaptic potential regulation of grooming behavior ureteric bud development cell adhesion protein localization regulation of protein localization nucleotide phosphorylation tissue morphogenesis positive regulation of multicellular organism growth positive regulation of developmental growth neuronal ion channel clustering locomotory behavior response to cocaine regulation of neuronal synaptic plasticity embryo development cell-cell adhesion neurotransmitter receptor localization to postsynaptic specialization membrane
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-56756dfb8b-mgmkt:80/100.66.75.22:80.
          git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.1-2025.06.25-1.0