Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Sodium Channel Pan Antibodies

          Antibodies Inc

          Pan-Nav1 Sodium Channel Monoclonal Antibody (N419/40), FL650

          View all (17) Sodium Channel Pan antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Pan-Nav1 Sodium Channel Monoclonal Antibody (N419/40), FL650

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          Pan-Nav1 Sodium Channel Monoclonal Antibody (N419/40), FL650

          Product Details

          75-405-FL650

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Frozen) (IHC (F))

          1:500
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          N419/40

          Immunogen

          Synthetic peptide amino acids 1501-1518 of human Nav1.1 (accession number P35498).
          View immunogen

          Conjugate

          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation

          Excitation/Emission Max

          655/676 nm

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Dry ice (international)

          Product Specific Information

          For maximum recovery of product, centrifuge the vial prior to removing the cap.

          Sequence of this protein is as follows: TEEQKKYYNA MKKLGSKK

          Target Information

          Voltage-gated sodium channels (NaV) are essential for the generation of action potentials and for cell excitability. NaV channels are activated in response to depolarization and selectively allow flow of Na+ ions. To date, nine NaV alpha subunits have been cloned and named NaV1.1-NaV1.9. The NaV channels are classified into two groups according to their sensitivity to tetrodotoxin (TTX): TTX-sensitive (NaV1.1, NaV1.2, NaV1.3, NaV1.4, NaV1.6 and NaV1.7) and TTX-resistant (NaV1.5, NaV1.8 and NaV1.9). Mammalian sodium channels are heterotrimers, composed of a central, pore-forming alpha subunit and two auxiliary beta subunits. The expression of the alpha subunit isoform is developmentally regulated and tissue specific. Sodium channels in the adult central nervous system and heart contain beta 1 through beta 4 subunits, whereas sodium channels in adult skeletal muscle have only the beta 1 subunit.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 3-Feb; alternative product; ataxia 3; brain III voltage-gated sodium channel; cardiac tetrodotoxin-insensitive voltage-dependent sodium channel alpha subunit; CTC-264K15.6; HBSC II; hH1; hNa6/Scn8a voltage-gated sodium channel; hNaN; hNE-Na; mH1; Mu-1; Na+ channel; NaCh6; NaN; Neuroendocrine sodium channel; Peripheral nerve protein type 4; Peripheral nerve sodium channel 3; Peripheral nerve sodium channel 5; Peripheral sodium channel 1; PN1; PN3; PN4; PN5; RIIA sodium channel protein; Sensory neuron sodium channel; Sensory neuron sodium channel 2; skeletal muscle sodium channel alpha subunit; skeletal muscle voltage-dependent sodium channel type IV alpha subunit; SkM1; sodium channel 25; Sodium channel 6; sodium channel alpha-subunit; Sodium channel protein brain I subunit alpha; Sodium channel protein brain II subunit alpha; Sodium channel protein brain III subunit alpha; Sodium channel protein cardiac muscle subunit alpha; Sodium channel protein skeletal muscle subunit alpha; Sodium channel protein type 1 subunit alpha; Sodium channel protein type 10 subunit alpha; Sodium channel protein type 11 subunit alpha; Sodium channel protein type 2 subunit alpha; Sodium channel protein type 3 subunit alpha; Sodium channel protein type 4 subunit alpha; Sodium channel protein type 5 subunit alpha; Sodium channel protein type 8 subunit alpha; Sodium channel protein type 9 subunit alpha; Sodium channel protein type I subunit alpha; Sodium channel protein type II subunit alpha; Sodium channel protein type III subunit alpha; Sodium channel protein type IV subunit alpha; Sodium channel protein type IX subunit alpha; Sodium channel protein type V subunit alpha; Sodium channel protein type VIII subunit alpha; Sodium channel protein type X subunit alpha; Sodium channel protein type XI subunit alpha; sodium channel protein, brain I alpha subunit; sodium channel protein, brain I subunit alpha; sodium channel protein, brain II subunit alpha; sodium channel protein, brain III subunit alpha; sodium channel protein, brain type 2 alpha subunit; sodium channel type X alpha polypeptide; sodium channel voltage gated type 1 alpha subunit; sodium channel voltage-gated type 11 alpha polypeptide; sodium channel voltage-gated type 4 alpha polypeptide; sodium channel voltage-gated type I alpha polypeptide; sodium channel voltage-gated type II alpha polypeptide; Sodium channel voltage-gated type IV alpha polypeptide; sodium channel voltage-gated type V alpha polypeptide; sodium channel voltage-gated type VIII alpha polypeptide; sodium channel voltage-gated type X alpha polypeptide; sodium channel voltage-gated type XI alpha polypeptide; sodium channel, voltage gated, type I alpha subunit; sodium channel, voltage gated, type II alpha subunit; sodium channel, voltage gated, type III alpha subunit; sodium channel, voltage gated, type IV alpha subunit; sodium channel, voltage gated, type IX alpha subunit; sodium channel, voltage gated, type V alpha subunit; sodium channel, voltage gated, type VIII alpha subunit; sodium channel, voltage gated, type VIII, alpha subunit; sodium channel, voltage gated, type X alpha subunit; sodium channel, voltage gated, type XI alpha subunit; sodium channel, voltage-gated, type 1, alpha polypeptide; sodium channel, voltage-gated, type 10, alpha polypeptide; sodium channel, voltage-gated, type 11, alpha polypeptide; sodium channel, voltage-gated, type 2, alpha 1 polypeptide; sodium channel, voltage-gated, type 2, alpha 1 subunit; sodium channel, voltage-gated, type 3, alpha subunit; sodium channel, voltage-gated, type 4, alpha polypeptide; sodium channel, voltage-gated, type 4, alpha subunit; sodium channel, voltage-gated, type 5, alpha subunit; sodium channel, voltage-gated, type 8, alpha polypeptide; sodium channel, voltage-gated, type 8, alpha subunit; sodium channel, voltage-gated, type 9, alpha polypeptide; sodium channel, voltage-gated, type I, alpha polypeptide; sodium channel, voltage-gated, type I, alpha subunit; sodium channel, voltage-gated, type II, alpha 1; sodium channel, voltage-gated, type II, alpha 1 polypeptide; sodium channel, voltage-gated, type II, alpha 2 polypeptide; sodium channel, voltage-gated, type II, alpha polypeptide 1; sodium channel, voltage-gated, type II, alpha subunit; sodium channel, voltage-gated, type III, alpha polypeptide; sodium channel, voltage-gated, type IV, alpha polypeptide; sodium channel, voltage-gated, type IV, alpha subunit; sodium channel, voltage-gated, type IX, alpha polypeptide; sodium channel, voltage-gated, type IX, alpha subunit; sodium channel, voltage-gated, type V, alpha polypeptide; sodium channel, voltage-gated, type V, alpha subunit; sodium channel, voltage-gated, type VIII, alpha polypeptide; sodium channel, voltage-gated, type X, alpha polypeptide; sodium channel, voltage-gated, type X, alpha subunit; sodium channel, voltage-gated, type XI, alpha polypeptide; sodium channel, voltage-gated, type XI, alpha subunit; sodium channel, voltage-gated, type XII, alpha polypeptide; sodium channel, voltage-gated, type11, alpha polypeptide; SP-19; voltage gated sodium channel; voltage-gated sodium channel alpha subunit Nav1.7; voltage-gated sodium channel Nav1.5c; voltage-gated sodium channel NAV1.9b; voltage-gated sodium channel subtype II; Voltage-gated sodium channel subtype III; Voltage-gated sodium channel subunit alpha Nav1.1; Voltage-gated sodium channel subunit alpha Nav1.2; Voltage-gated sodium channel subunit alpha Nav1.3; Voltage-gated sodium channel subunit alpha Nav1.4; Voltage-gated sodium channel subunit alpha Nav1.5; Voltage-gated sodium channel subunit alpha Nav1.6; Voltage-gated sodium channel subunit alpha Nav1.7; Voltage-gated sodium channel subunit alpha Nav1.8; Voltage-gated sodium channel subunit alpha Nav1.9; voltage-gated sodium channel type II alpha subunit; voltage-gated sodium channel type VIII alpha protein

          View more View less

          Gene Aliases: 6430408L10; A230052E19Rik; AI853486; B230332M13; BFIC3; BFIS3; BFNIS; C630029C19Rik; CDCD2; CERIII; CIAT; CMD1E; CMPD2; CMS16; dmu; EIEE11; EIEE13; EIEE6; ETHA; FEB3; FEB3A; FEB3B; FEPS2; FEPS3; FHM3; GEFSP2; GEFSP7; Gm1000; HB1; HB2; HBA; HBBD; HBSCI; HBSCII; HH1; HOKPP2; hPN3; HSAN2D; HSAN7; HYKPP; HYPP; ICCD; IVF; KIAA1356; Kiaa4197; LQT3; MED; mH1; mH2; microI; mKIAA4197; mnd-2; mnd2; Na(v)1.2; Na(V)1.4; Na(V)1.8; NAC1; NAC1A; NAC2; NAC3; NaCh6; NachII; NaN; NaT; Nav1.1; Nav1.2; Nav1.3; Nav1.4; Nav1.5; Nav1.5c; Nav1.6; Nav1.7; Nav1.8; NAV1.9; Nbna1; NCHVS; NE-NA; NENA; nmf2; nmf335; nmf58; NSS2; nur14; PFHB1; PN1; PN3; PN4; PN5; RII/RIIA; RNSCPIIR; SCAL; SCIII; SCN; SCN1; SCN10A; SCN11A; SCN12A; SCN1A; SCN2A; SCN2A1; SCN2A2; SCN3A; SCN4A; SCN5A; SCN8A; SCN9A; ScpII; seal; SFNP; SkM1; SkM2; SMEI; SNS; SNS-2; SNS2; SSS1; VF1

          View more View less

          UniProt ID: (Human) Q9Y5Y9, (Human) Q9UI33, (Human) P35498, (Human) Q99250, (Human) Q9NY46, (Human) P35499, (Human) E9PEK2, (Human) Q9UQD0, (Human) Q15858, (Rat) Q62968, (Mouse) Q6QIY3, (Rat) O88457, (Mouse) Q9R053, (Rat) P04774, (Mouse) A2APX8, (Rat) P04775, (Mouse) B1AWN6, (Mouse) A2ASI5, (Rat) P08104, (Mouse) Q9ER60, (Rat) P15390, (Rat) P15389, (Mouse) Q9JJV9, (Mouse) Q9WTU3, (Rat) O88420, (Mouse) Q62205, (Rat) O08562

          View more View less

          Entrez Gene ID: (Human) 6336, (Human) 11280, (Human) 6323, (Human) 6326, (Human) 6328, (Human) 6329, (Human) 6331, (Human) 6334, (Human) 6335, (Rat) 29571, (Mouse) 20264, (Rat) 29701, (Mouse) 24046, (Rat) 81574, (Mouse) 20265, (Rat) 24766, (Mouse) 110876, (Mouse) 20269, (Rat) 497770, (Mouse) 110880, (Rat) 25722, (Rat) 25665, (Mouse) 20271, (Mouse) 20273, (Rat) 29710, (Mouse) 20274, (Rat) 78956

          View more View less

          Function(s)
          voltage-gated sodium channel activity ion channel binding sodium ion binding calmodulin binding protein binding fibroblast growth factor binding enzyme binding protein kinase binding protein domain specific binding ankyrin binding ubiquitin protein ligase binding nitric-oxide synthase binding voltage-gated sodium channel activity involved in cardiac muscle cell action potential voltage-gated sodium channel activity involved in AV node cell action potential voltage-gated sodium channel activity involved in bundle of His cell action potential voltage-gated sodium channel activity involved in Purkinje myocyte action potential voltage-gated sodium channel activity involved in SA node cell action potential scaffold protein binding ATP binding ion channel activity voltage-gated ion channel activity sodium channel activity leucine zipper domain binding nucleotide binding voltage-gated ion channel
          Process(es)
          regulation of heart rate sensory perception neuronal action potential sensory perception of pain regulation of ion transmembrane transport sodium ion transmembrane transport odontogenesis of dentin-containing tooth regulation of cardiac muscle contraction regulation of postsynaptic membrane potential regulation of atrial cardiac muscle cell membrane depolarization membrane depolarization during action potential AV node cell action potential bundle of His cell action potential sodium ion transport response to drug regulation of sensory perception of pain positive regulation of defense response to virus by host adult walking behavior neuronal action potential propagation neuromuscular process controlling posture xenophagy intrinsic apoptotic signaling pathway in response to osmotic stress myelination neuron apoptotic process nervous system development response to wounding response to pyrethroid cellular response to antibiotic potassium ion transport muscle contraction choline transport cardiac ventricle development brainstem development positive regulation of sodium ion transport response to denervation involved in regulation of muscle adaptation telencephalon development cerebellum development positive regulation of action potential positive regulation of epithelial cell proliferation membrane depolarization cardiac muscle contraction regulation of ventricular cardiac muscle cell membrane repolarization regulation of atrial cardiac muscle cell membrane repolarization regulation of ventricular cardiac muscle cell membrane depolarization cardiac conduction cellular response to calcium ion cardiac muscle cell action potential involved in contraction regulation of cardiac muscle cell contraction ventricular cardiac muscle cell action potential membrane depolarization during cardiac muscle cell action potential SA node cell action potential membrane depolarization during AV node cell action potential membrane depolarization during SA node cell action potential membrane depolarization during Purkinje myocyte cell action potential membrane depolarization during bundle of His cell action potential AV node cell to bundle of His cell communication regulation of heart rate by cardiac conduction regulation of sodium ion transmembrane transport peripheral nervous system development inflammatory response response to toxic substance post-embryonic development behavioral response to pain bundle of His cell to Purkinje myocyte communication transport ion transport transmembrane transport action potential regulation of membrane potential cation transport positive regulation of heart rate response to organic cyclic compound SA node cell to atrial cardiac muscle cell communication muscle organ development sensory perception of sound locomotory behavior adult locomotory behavior neuromuscular process rhythmic excitation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0