Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Pan-PTPR Antibodies

          Antibodies Inc

          Pan-PTPR (Receptor-Type Tyrosine-Protein Phosphatase) Monoclonal Antibody (N165/43), FL650

          View all (5) Pan-PTPR antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Pan-PTPR (Receptor-Type Tyrosine-Protein Phosphatase) Monoclonal Antibody (N165/43), FL650

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          Pan-PTPR (Receptor-Type Tyrosine-Protein Phosphatase) Monoclonal Antibody (N165/43), FL650

          Product Details

          75-194-FL650

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          N165/43

          Immunogen

          Fusion protein amino acids 1315-1607 (cytoplasmic C-terminus) of human LAR (accession number P10586) produced recombinantly in E. Coli.
          3D Epitope / Immunogen

          Conjugate

          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation

          Excitation/Emission Max

          655/676 nm

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Dry ice (international)

          Product Specific Information

          For maximum recovery of product, centrifuge the vial prior to removing the cap.

          Target Information

          Pan-PTPR refers to the collective group of Protein Tyrosine Phosphatase Receptor (PTPR) family members, which are enzymes that dephosphorylate tyrosine residues on proteins. These receptors play key roles in regulating a variety of cellular processes, including cell growth, differentiation, mitotic cycle, and oncogenic transformation. Their dysregulation is often associated with various diseases, including cancer and autoimmune disorders.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: LAR; LCA-homolog; leukocyte antigen-related (LAR) PTP receptor; leukocyte antigen-related tyrosine phosphatase; leukocyte common antigen related; leukocyte common antigen-related protein-tyrosine phosphatase 2; leukocyte common antigen-related proten-tyrosine phosphatase 2; protein tyrosine phosphatase PTPsigma; protein tyrosine phosphatase, receptor type, D; protein tyrosine phosphatase, receptor type, delta polypeptide; protein tyrosine phosphatase, receptor type, F polypeptide; rceptor-type tyrosine-protein phosphatase delta; receptor-linked protein-tyrosine phosphatase LAR; Receptor-type tyrosine-protein phosphatase sigma; tissue-specific protein tyrosine phosphatase; unnamed protein product

          View more View less

          Gene Aliases: 1110002J03Rik; 3000002J10Rik; B230219D21Rik; BNAH2; HPTP; HPTPD; HPTPDELTA; LAR; LARFN5C; Larptp2; LARS; PTP; PTP-NU3; PTP-sigma; PTPD; PTPNU-3; Ptprd; PTPSIGMA; Ptpsigma.; Ptpt9; R-PTP-delta; R-PTP-S; R-PTP-sigma; RPTP-LAR; Rptpd; RPTPDELTA; RPTPsigma

          View more View less

          UniProt ID: (Human) P23468, (Human) P10586, (Human) Q13332, (Mouse) Q64487, (Mouse) A2A8L5, (Rat) Q64604, (Mouse) B0V2N1, (Rat) Q64605

          View more View less

          Entrez Gene ID: (Human) 5789, (Human) 5792, (Human) 5802, (Rat) 313278, (Mouse) 19266, (Mouse) 19268, (Rat) 360406, (Mouse) 19280, (Rat) 25529

          View more View less

          Function(s)
          protein tyrosine phosphatase activity transmembrane receptor protein tyrosine phosphatase activity receptor binding protein binding cell adhesion molecule binding phosphoprotein phosphatase activity chondroitin sulfate proteoglycan binding macromolecular complex binding heparin binding chondroitin sulfate binding heparan sulfate proteoglycan binding insulin receptor binding receptor tyrosine kinase binding phosphate ion binding
          Process(es)
          heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules signal transduction nervous system development regulation of neuron projection development neuron differentiation cellular response to platelet-derived growth factor stimulus negative regulation of JAK-STAT cascade positive regulation of dendrite morphogenesis regulation of immune response modulation of synaptic transmission positive regulation of synapse assembly positive regulation of dendritic spine morphogenesis cellular response to hypoxia presynaptic membrane assembly presynapse assembly regulation of postsynaptic density assembly trans-synaptic signaling trans-synaptic signaling by trans-synaptic complex synaptic membrane adhesion negative regulation of cytokine-mediated signaling pathway protein dephosphorylation homophilic cell adhesion via plasma membrane adhesion molecules negative regulation of cell proliferation cell migration neuron projection regeneration negative regulation of cell projection organization peptidyl-tyrosine dephosphorylation negative regulation of epidermal growth factor receptor signaling pathway positive regulation of apoptotic process positive regulation of neuron apoptotic process negative regulation of insulin receptor signaling pathway regulation of axon regeneration regulation of synapse structure or activity negative regulation of neurotrophin TRK receptor signaling pathway regulation of postsynapse organization negative regulation of receptor binding negative regulation of neuron projection development spinal cord development cerebellum development hippocampus development cerebral cortex development corpus callosum development negative regulation of axon extension negative regulation of interferon-alpha production negative regulation of interferon-beta production negative regulation of toll-like receptor 9 signaling pathway negative regulation of collateral sprouting negative regulation of axon regeneration synapse organization regulation of dendritic spine development negative regulation of dendritic spine development establishment of endothelial intestinal barrier cell adhesion transmembrane receptor protein tyrosine phosphatase signaling pathway phosphate-containing compound metabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7bf6cff7cf-ct9mq:80/100.66.77.168:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0