Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Isotype Controls ›
          • Phalloidin Isotype Controls
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.

          Invitrogen

          Phalloidin, DyLight™ 550

          View all Phalloidin Controls

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support

          Cite Phalloidin, DyLight™ 550

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Testing Data (8)
          Phalloidin Control in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phalloidin Control in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phalloidin Control (21835) in ICC/IF

          Immunofluorescent analysis of Phalloidin (orange) and alpha-Tubulin (green) in NIH 3T3 cells. Formalin fixed cells were permeabilized with 0.1% Triton X-100 in PBS for 10 minutes at room temperature and blocked with 2% BSA (Product # 37525) in PBS + 0.1% Triton X-100 for 30 minutes at room temperature. Cells were probed with an alpha-Tubulin monoclonal antibody (Product # MA1-19162) at a dilution of 1:1000 ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phalloidin Control in Immunocytochemistry (ICC/IF)
          Phalloidin Control in Immunocytochemistry (ICC/IF)
          Phalloidin Control in Immunocytochemistry (ICC/IF)
          Phalloidin Control in Immunocytochemistry (ICC/IF)
          Phalloidin Control in Immunocytochemistry (ICC/IF)
          Phalloidin Control in Immunocytochemistry (ICC/IF)
          Phalloidin Control in Immunocytochemistry (ICC/IF)
          Phalloidin Control in Immunocytochemistry (ICC/IF)

          Product Details

          21835

          Applications
          Tested Dilution

          Immunohistochemistry (IHC)

          Assay-dependent

          Immunocytochemistry (ICC/IF)

          1-5 units/mL
          Product Specifications

          Type

          Control

          Conjugate

          DyLight™ 550 DyLight™ 550 DyLight™ 550

          Excitation/Emission Max

          556/570 nm View spectra spectra

          Form

          Lyophilized

          Concentration

          300 U/mL

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          RRID

          AB_2532157

          Product Specific Information

          Format: 300 units of lyophilized DyLight 550-Phalloidin (300 units/mL) in methanol. Prepare stock solution by adding 1000 µL of pure methanol to the vial and gently mix.

          21835 has been successfully used in immunofluorescence and immunohistochemistry. DyLight 550-Phalloidin has an excitation/emission of 557/573 nm and molecular weight of 1452.68 g/mole.

          DyLight 550-Phalloidin Stock Solution is prepared in methanol, and is also soluble in DMF, or DMSO. The unused Stock Solution should be promptly stored at -20°C in a foil pouch with desiccant, and protected from light. The stock solutions are stable for at least one year. Staining cells with some of the DyLight Phalloidin conjugates may require the use of a higher concentration of the phalloidin conjugate; therefore to minimize the amount of methanol added to the cells, the vial contents could be dissolved in 0.5 mL methanol to yield a final concentration of 600units/mL.

          Working solution of 1unit/mL (per 96-well plate) can be prepared by diluting 20 µL of the Stock Solution in 5.98 mL of PBS and mixing well. Typical staining procedure adds 50 µL of Working Solution (i.e. 1 to 5 unit/mL) to a each well. Incubate cells in the dark for 30 minutes at room temperature (optimal staining times varies from 10 minutes to 3 hours depending on cell type). Aspirate and wash cells three times in PBS after incubation. If desired, probe with specific primary antibodies followed by secondary antibodies conjugated to any compatible fluorophores before using DyLight 550-Phalloidin conjugate.

          Target Information

          Phalloidin is a bicyclic peptide that belongs to a family of toxins isolated from the deadly Amanita phalloides 'death cap' mushroom and is commonly used as a counterstain (similar to DAPI or Hoechst) in cell biology and histology imaging applications to selectively label F-actin in fixed cells, permeabilized cells, and cell-free experiments. Labeled phalloidin conjugates have similar affinity for both large and small filaments and bind in a stoichiometric ratio of about one phallotoxin per actin subunit in both muscle and non-muscle cells. Phalloidins reportedly do not bind to monomeric G-actin, unlike some antibodies against actin. The dynamics of the actin polymerization in cells are important for a variety of cellular processes from cell motility to cell shape, from muscular contraction to cytokinesis, and more.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          View product citations for antibody 21835 on CiteAb

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline