MA1-10739 detects Phenytoin.
MA1-10739 has been successfully used in ELISA applications.
The MA1-10739 immunogen is Phenytoin, conjugated to KLH.
Phenytoin sodium is a commonly used antiepileptic. Phenytoin (fen-i-TOE-in) acts to suppress the abnormal brain activity seen in seizure by reducing electrical conductance among brain cells by stabilizing the inactive state of voltage-gated sodium channels. Aside from seizures, it is an option in the treatment of trigeminal neuralgia in the event that carbamazepine is deemed inappropriate.
Protein Aliases: Dilantin; Epanutin; Infatabs; Kapseals; Phenytek