Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
This antibody detects endogenous protein at a molecular weight of 35 and 39 kDa.
Purity is >95% by SDS-PAGE.
Aurora kinases belong to a highly conserved family of mitotic serine/threonine kinases with three members identified among mammals: Aurora A, B, and C. Studies on the temporal expression pattern and subcellular localization of Aurora kinases in mitotic cells suggest an association with mitotic structure. Aurora kinase functional influences span from G2 phase to cytokinesis and may be involved in key cell cycle events such as centrosome duplication, chromosome bi-orientation and segregation, cleavage furrow positioning, and ingression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AIM-1; ARK-2; ARK-3; Aurora 1; Aurora 3; aurora B; Aurora kinase B; aurora kinase B-Sv1; aurora kinase B-Sv2; Aurora kinase C; Aurora- and IPL1-like midbody-associated protein 1; aurora-1; aurora-B; aurora-related kinase 2; aurora-related kinase 3; Aurora/IPL1-related kinase 2; Aurora/IPL1-related kinase 3; Aurora/IPL1/Eg2 protein 1; Aurora/IPL1/Eg2 protein 2; epididymis secretory protein Li 90; protein phosphatase 1, regulatory subunit 48; serine/threonine kinase 12; serine/threonine kinase 13 (aurora/IPL-like); serine/threonine kinase 13 (aurora/IPL1-like); serine/threonine kinase 5; Serine/threonine-protein kinase 12; Serine/threonine-protein kinase 13; Serine/threonine-protein kinase 5; Serine/threonine-protein kinase aurora-B; Serine/threonine-protein kinase aurora-C; STK-1
Gene Aliases: AIE1; AIE2; AIK2; AIK3; AIM-1; AIM1; AIRK2; AIRK3; AL022959; ARK-2; ARK-3; ARK2; ARK3; AurB; AurC; AURKB; aurkb-sv1; aurkb-sv2; AURKC; aurora-C; HEL-S-90; IAK3; IPL1; PPP1R48; SPGF5; STK-1; STK1; STK12; STK13; STK5
UniProt ID: (Human) Q96DV5, (Human) Q9UQB9, (Mouse) O70126, (Rat) O55099, (Mouse) O88445
Entrez Gene ID: (Human) 9212, (Human) 6795, (Mouse) 20877, (Rat) 114592, (Mouse) 20871, (Rat) 292554
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support