Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CaMKII beta/gamma/delta Antibodies

          Invitrogen

          Phospho-CaMKII beta/gamma/delta (Thr287) Polyclonal Antibody

          Advanced Verification
          9 Published Figures
          19 References
          View all (4) CaMKII beta/gamma/delta antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-CaMKII beta/gamma/delta (Thr287) Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (10)
          • Published Figures (9)
          • Advanced Verification (1)
          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 20

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody (PA5-37833) in IHC (P)

          Immunohistochemistry analysis of Phospho-CaMKII beta/gamma/delta (Thr287) on paraffin-embedded human lung tissue using Phospho-CaMKII beta/gamma/delta (Thr287) Polyclonal Antibody (Product # PA5-37833) at dilution 1:300 (room temperature, 50 min). Picture B: DAPI (blue) 10 min. Picture A: Phospho-CaMKII beta/gamma/delta (Thr287). Picture B: DAPI. Picture C: merge of A+B. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody in Western Blot (WB)
          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody in Western Blot (WB)
          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody in Western Blot (WB)
          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody in Western Blot (WB)
          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody in Western Blot (WB)
          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody in Western Blot (WB)
          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody in Western Blot (WB)
          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody in Western Blot (WB)
          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody in Western Blot (WB)
          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody in Western Blot (WB)
          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody in Western Blot (WB)
          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody in Western Blot (WB)
          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody in Western Blot (WB)
          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody in Western Blot (WB)
          Phospho-CaMKII beta/gamma/delta (Thr287) Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-CaMKII beta/gamma/delta (Thr287) Polyclonal Antibody

          Product Details

          PA5-37833

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 17 publications 17 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:300
          -

          Flow Cytometry (Flow)

          -
          View 1 publication 1 publication

          ELISA (ELISA)

          1:5,000
          -

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Pig, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Peptide sequence around phosphorylation site of Threonine 287 (Q-E-T(p)-V-E) derived from Human CaMK2- beta/ gamma/ delta
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol, 0.5% BSA

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2554441

          Product Specific Information

          A suggested positive control for Western blot is rat brain cells.

          Target Information

          The Ca2+/calmodulin-dependent protein kinases (CaM kinases) comprise a structurally related subfamily of serine/threonine kinases which include CaMKI, CaMKII and CaMKIV. CaMKII is an ubiquitously expressed serine/threonine protein kinase that is activated by Ca2+ and calmodulin (CaM) and has been implicated in regulation of the cell cycle and transcription. There are four CaMKII isozymes, designated alpha, beta, gamma and delta, which may or may not be coexpressed in the same tissue type. CaMKIV is stimulated by Ca2+ and CaM but also requires phosphorylation by a CaMK for full activation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: alternatively spliced; association domain; beta M CaM kinase II; brain type II Ca2+/calmodulin-dependent protein kinase beta subunit; Ca++/calmodulin-dependent protein kinase 2 delta subunit; Ca++/calmodulin-dependent protein kinase II beta subunit; Ca++/calmodulin-dependent protein kinase II, delta subunit; Ca+2/calmodulin-dependent protein kinase II gamma; Ca2+/calmodulin-dependent protein kinase II; Calcium, calmodulin-dependent protein kinase; Calcium, calmodulin-dependent protein kinase II; Calcium, calmodulin-dependent protein kinase, type II; calcium/calmodulin-dependent protein kinase (CaM kinase) II beta; calcium/calmodulin-dependent protein kinase (CaM kinase) II delta; calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma; calcium/calmodulin-dependent protein kinase type II beta chain; calcium/calmodulin-dependent protein kinase type II delta chain; calcium/calmodulin-dependent protein kinase type II gamma chain; calcium/calmodulin-dependent protein kinase type II subunit gamma-like; calmodulin-dependent protein kinase II gamma subunit (EC 2.7.1.37); calmodulin-dependent protein kinase II-delta (EC 2.7.1.37); CaM kinase II; CaM kinase II beta subunit; CaM kinase II delta subunit; caM kinase II subunit beta; caM kinase II subunit delta; caM kinase II subunit gamma; CaM-kinase II beta chain; CaM-kinase II delta chain; caM-kinase II gamma chain; CaMK II; CaMK-II delta subunit; caMK-II subunit beta; caMK-II subunit delta; caMK-II subunit gamma; Camk2 gamma; proline rich calmodulin-dependent protein kinase; protein kinase homolog; unnamed protein product

          View more View less

          Gene Aliases: 2810011D23Rik; 8030469K03Rik; [d]-CaMKII; CAM2; CAMK; CaMK II; CAMK-II; CAMK1; CAMK2; CAMKB; CAMKD; CAMKG; Camki; CaMKII; CaMKIIbeta; Ck2b; MRD54; MRD59; RATCAMKI

          View more View less

          UniProt ID: (Human) Q13554, (Human) Q13557, (Human) Q13555, (Rat) P08413, (Mouse) P28652, (Rat) P15791, (Mouse) Q6PHZ2, (Mouse) Q923T9, (Rat) P11730

          View more View less

          Entrez Gene ID: (Human) 816, (Human) 817, (Human) 818, (Rat) 24245, (Mouse) 12323, (Rat) 24246, (Mouse) 108058, (Mouse) 12325, (Rat) 171140

          View more View less

          Function(s)
          protein kinase activity calmodulin-dependent protein kinase activity protein binding calmodulin binding ATP binding identical protein binding protein homodimerization activity protein serine kinase activity protein serine/threonine kinase activity sodium channel inhibitor activity titin binding ion channel binding calcium-dependent protein serine/threonine phosphatase activity protein kinase binding phospholipase binding structural constituent of postsynaptic actin cytoskeleton nitric-oxide synthase binding
          Process(es)
          regulation of cell growth regulation of the force of heart contraction regulation of membrane depolarization regulation of transcription from RNA polymerase II promoter protein phosphorylation regulation of heart contraction positive regulation of cardiac muscle hypertrophy regulation of cell communication by electrical coupling positive regulation of cardiac muscle cell apoptotic process regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion endoplasmic reticulum calcium ion homeostasis regulation of neuronal synaptic plasticity relaxation of cardiac muscle long-term synaptic potentiation regulation of ryanodine-sensitive calcium-release channel activity regulation of cellular localization cellular response to calcium ion cardiac muscle cell contraction regulation of heart rate by cardiac conduction regulation of cardiac muscle cell action potential regulation of cardiac muscle cell action potential involved in regulation of contraction regulation of cell communication by electrical coupling involved in cardiac conduction regulation of relaxation of cardiac muscle negative regulation of sodium ion transmembrane transport regulation of calcium ion transmembrane transport via high voltage-gated calcium channel regulation of protein localization to plasma membrane G1/S transition of mitotic cell cycle inhibitory G-protein coupled receptor phosphorylation calcium ion transport positive regulation of phospholipase activity positive regulation of neuron projection development peptidyl-serine phosphorylation regulation of synaptic transmission, cholinergic response to cadmium ion protein autophosphorylation regulation of long-term neuronal synaptic plasticity neuromuscular process controlling balance activation of meiosis involved in egg activation positive regulation of dendritic spine morphogenesis regulation of synapse maturation positive regulation of synapse maturation postsynaptic actin cytoskeleton organization hippocampal neuron apoptotic process cellular response to homocysteine response to psychosocial stress regulation of neuron migration positive regulation of apoptotic signaling pathway signal transduction regulation of skeletal muscle adaptation regulation of synapse structural plasticity regulation of calcium ion transport regulation of dendritic spine development response to hypoxia response to oxidative stress regulation of neuron projection development regulation of postsynaptic membrane neurotransmitter receptor levels regulation of sodium ion transport positive regulation of G2/M transition of mitotic cell cycle positive regulation of smooth muscle cell migration cellular potassium ion homeostasis positive regulation of Rac protein signal transduction cardiac muscle contraction cell growth involved in cardiac muscle cell development positive regulation of ERK1 and ERK2 cascade positive regulation of vascular smooth muscle cell proliferation positive regulation of DNA biosynthetic process insulin secretion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5d6d8bc979-v94d6:80/100.66.72.150:80.
          git-commit: e71199a02a6c1449b2ac9ded8316d5734e8aa2de
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.62.0-2026.11.45-1.0