Caveolae are specialized domains of the plasma membrane that are implicated in the sequestration of a variety of lipid and protein molecules. It has been suggested that these important cellular organelles have a pivotal role in such diverse biochemical processes as lipid metabolism, growth regulation, signal transduction, and apoptosis. Caveolin interacts with and regulates heterotrimeric G-proteins. Currently, there are three members of the caveolin multigene family which are known to encode 21-24 kDa integral membrane proteins that comprise the major structural component of the caveolar membrane in vivo. Caveolin-2 protein is abundantly expressed in fibroblasts and differentiated adipocytes, smooth and skeletal muscle, and endothelial cells. The expression of caveolin-1 is similar to that of caveolin-2 while caveolin-3 expression appears to be limited to muscle tissue types.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: caveolae; caveolae protein, 20-kD; caveolin 2 isoform a and b; Caveolin-2; MGC12294; Phospho-CAV2
Gene Aliases: AI447843; CAV; CAV2
UniProt ID: (Human) P51636, (Mouse) Q9WVC3, (Rat) Q2IBC5
Entrez Gene ID: (Human) 858, (Mouse) 12390, (Rat) 363425
Molecular Function:
G-protein modulator
enzyme modulator
membrane traffic protein
structural protein
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support