Invitrogen
Antibody target was verified by Cell Treatment to ensure the antibody binds to the antigen stated. View details.
Catalog #:700012
Quantity:
Usually Ships In 1 business day
| TESTED APPLICATIONS | DILUTION |
|---|---|
| ChIP assay (ChIP) | 1 ul |
| ELISA (ELISA) | Assay-dependent |
| Immunocytochemistry (ICC) | 5 µg/ml |
| Immunofluorescence (IF) | 5 µg/ml |
| Immunohistochemistry (Paraffin) (IHC (P)) | 5 ug/ml |
| Western Blot (WB) | 1 µg/ml |
| PUBLISHED APPLICATIONS | |
|---|---|
| Western Blot (WB) | See 2 publications below |
| Tested Species reactivity | Dog, Human, Rat |
| Published species reactivity | Human Mouse |
| Host / Isotype | Rabbit / IgG |
| Class | Monoclonal |
| Type | Antibody |
| Clone | 15H10L7 |
| Immunogen | A peptide corresponding to amino acids 198-208 and 181-191 of P27361 and P28482, respectively. |
| Conjugate | Unconjugated |
| Form | Liquid |
| Concentration | 0.5 mg/ml |
| Purification | Protein A |
| Storage buffer | PBS |
| Contains | 0.09% sodium azide |
| Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
| RRID | AB_2532267 |
700012 has successfully been used in western blot to detect phospho-ERK1/2 in TPA-treated HeLa, NRK and MDCK cell lysates.
This antibody is predicted to react with bovine, chicken, chimpanzee, mouse, rat, Xenopus and zebrafish based on sequence homology.
ABfinity™ recombinant antibodies are rabbit monoclonal antibodies, unmatched for producing superior results. ABfinity™ antibodies are developed by immunizing animals, screening for functionality, cloning the immunogen-specific antibody genes into high-level mammalian expression vectors, produced on a large scale, and purified with Protein A.
ABfinity™ monoclonal antibodies resemble rabbit monoclonals isolated from serum or produced by hybridomas, but demonstrate greater specificity and sensitivity. Because ABfinity™ recombinant antibodies are derived from cloned DNA sequences of the heavy and light antibody chains, they are not susceptible to cell-line drift or lot-to-lot variation, thus allowing for peak specificity and performance.
Intact IgG appears on a non-reducing gel as ~150 kDa band and upon reduction generating a ~25 kDa light chain band and a ~50 kDa heavy chain.
Mitogen activated protein kinase (MAPK) belongs to a family of serine/threonine kinases that are activated upon exposure of cells to a variety of stimuli including mitogens, hormones, growth factors, cytokines and bioactive peptides. There are two closely related isoforms of MAPK, with apparent molecular weights of 44 kDa and 42 kDa. They are abundantly expressed in neurons of the mature central nervous system. Activated MAPK plays a role in cell division and differentiation, and regulation of the cytoskeleton.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ERK-1; ERK-2; ERK1; ERK2; ERT1; ERT2; Extracellular signal-regulated kinase 1; Extracellular signal-regulated kinase 2; extracellular signal-related kinase 1; extracellular-signal-regulated kinase 2; Insulin-stimulated MAP2 kinase; MAP kinase 1; MAP kinase 2; MAP kinase 3; MAP kinase isoform p42; MAP kinase isoform p44; MAPK 1; MAPK 2; MAPK 3; MAPK1; MAPK3; Microtubule-associated protein 2 kinase; mitogen-activated 3; Mitogen-activated protein kinase 1; Mitogen-activated protein kinase 2; Mitogen-activated protein kinase 3; MNK1; p42-MAPK; p44-ERK1; p44-MAPK; PRKM1; PRKM2; PRKM3; protein tyrosine kinase ERK2
Gene Aliases: ERK; ERK-1; ERK-2; ERK1; ERK2; ERT1; ERT2; Esrk1; HS44KDAP; HUMKER1A; Mapk; MAPK1; MAPK2; MAPK3; MNK1; p38; p40; p41; p41mapk; p42-MAPK; P42MAPK; p44; p44-ERK1; p44-MAPK; P44ERK1; P44MAPK; PRKM1; PRKM2; PRKM3
UniProt ID: (Human) P28482, (Human) P27361, (Rat) P63086, (Rat) P21708
Entrez Gene ID: (Dog) 477575, (Human) 5594, (Human) 5595, (Dog) 100855877, (Rat) 116590, (Rat) 50689
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.