Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • CDMO and Clinical Trial Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Instrument Management

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Ezrin Antibodies

          Invitrogen

          Phospho-Ezrin (Tyr353) Polyclonal Antibody

          View all (33) Ezrin antibodies
          Datasheet
          Protocols
          Questions & Answers
          Tech Support

          Cite Phospho-Ezrin (Tyr353) Polyclonal Antibody

          Datasheet
          Protocols
          Questions & Answers
          Tech Support

          Product Details

          PA5-17683

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Dog, Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic phosphopeptide corresponding to residues surrounding pTyr353 of human Ezrin
          View immunogen

          Conjugate

          Unconjugated

          Form

          Liquid

          Purification

          Antigen affinity chromatography

          Storage buffer

          0.01M HEPES, pH 7.5, with 0.15M NaCl, 100µg/mL BSA, 50% glycerol

          Contains

          no preservative

          Storage conditions

          -20°C

          RRID

          AB_10979996

          Product Specific Information

          It is not recommended to aliquot this antibody.

          This antibody is not cross-reactive with phosphorylated Moesin or Radixin.

          Target Information

          The cytoplasmic peripheral membrane protein encoded by this gene functions as a protein-tyrosine kinase substrate in microvilli. As a member of the ERM protein family, this protein serves as an intermediate between the plasma membrane and the actin cytoskeleton. This protein plays a key role in cell surface structure adhesion, migration and organization, and it has been implicated in various human cancers. A pseudogene located on chromosome 3 has been identified for this gene. Alternatively spliced variants have also been described for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Cytovillin; cytovillin 2; DKFZp762H157; epididymis secretory protein Li 105; EZRI; Ezrin; FLJ26216; MGC1584; p81; villin 2 (ezrin); Villin-2

          View more View less

          Gene Aliases: CVIL; CVL; EZR; HEL-S-105; VIL2

          View more View less

          UniProt ID: (Human) P15311

          View more View less

          Entrez Gene ID: (Human) 7430, (Dog) 484056

          View more View less

          Molecular Function: actin or actin-binding cytoskeletal protein

          Function(s)
          negative regulation of transcription from RNA polymerase II promoter intestinal D-glucose absorption sphingosine-1-phosphate signaling pathway cytoskeletal anchoring at plasma membrane leukocyte cell-cell adhesion axon guidance regulation of cell shape regulation of cell size positive regulation of gene expression protein kinase A signaling gland morphogenesis membrane to membrane docking microvillus assembly astral microtubule organization actin cytoskeleton reorganization receptor internalization regulation of microvillus length regulation of actin cytoskeleton organization cellular protein complex localization positive regulation of multicellular organism growth cortical microtubule organization establishment of epithelial cell apical/basal polarity filopodium assembly phosphatidylinositol-mediated signaling positive regulation of protein secretion negative regulation of T cell receptor signaling pathway actin filament bundle assembly establishment of centrosome localization establishment of endothelial barrier negative regulation of ERK1 and ERK2 cascade cellular response to cAMP protein localization to cell cortex establishment of protein localization to plasma membrane positive regulation of establishment of protein localization to plasma membrane negative regulation of interleukin-2 secretion regulation of NIK/NF-kappaB signaling regulation of organelle assembly terminal web assembly positive regulation of protein localization to early endosome positive regulation of cellular protein catabolic process negative regulation of p38MAPK cascade positive regulation of early endosome to late endosome transport
          Process(es)
          actin binding protein binding microtubule binding protein domain specific binding protein kinase A catalytic subunit binding protein kinase A regulatory subunit binding S100 protein binding poly(A) RNA binding cell adhesion molecule binding actin filament binding ATPase binding cadherin binding involved in cell-cell adhesion
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © Copyright 2006-2023 Thermo Fisher Scientific Inc. All rights reserved
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-srch-2-prod-blue:8080/10.253.226.4:8080.
          git-commit: 21c286debe71d856c9209ca25c11ffa6178afad8
          git-url: git@github.com:thermofisher/magellan-core.git
          git-branch: origin/release/versionUpgradeChanges