Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OPA1-03888 detects phospho-FAK (pTyr576) from human samples. The mouse, rat, chicken, and, Xenopus laevis proteins are 100% conserved but have not been tested at this time.
OPA1-03888 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~125 kDa protein representing phospho-FAK (pTyr576).
OPA1-03888 immunizing phosphopeptide is derived from the region of FAK that contains tyrosine 576.
Focal Adhesion Kinase (FAK) is a 125 kDa non-receptor protein tyrosine kinase that acts as a substrate for Src and is a key element of integrin signaling. FAK plays an important role in cell spreading, differentiation, migration, cell death, and acceleration of the G1 to S phase transition of the cell cycle. Tyrosine 397 is the autophosphorylation site of FAK, and involved in its initial activation. This phosphorylated site binds Src family SH2 domains and the p85 subunit of PI3-Kinase, and activates cell migration and invasion. FAK has a central catalytic domain and a C-terminal tail that localizes it to focal adhesions, which are sites where cells attach to the extracellular matrix via surface integrin receptors. Increased FAK tyrosine phosphorylation occurs upon integrin engagement with fibronectin. Adhesion of murine NIH3T3 fibroblasts to fibronectin promotes association of the Grb2 adapter protein and c-Src PTK with FAK in vivo, and also results in activation of the ERK2 MAP kinase. In v-Src-transformed NIH3T3, the association of v-Src, Grb2, and Sos with FAK is independent of cell adhesion to fibronectin. In vitro the Grb2 SH2 domain binds directly to tyrosine-phosphorylated FAK, and the binding site has been identified as Tyr925 by site directed mutagenesis. Several transcript variants encoding different isoforms have been found for the FAK gene, but the full-length natures of only three of them have been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 2.7.10.2; FADK 1; FAK-related non-kinase polypeptide; Focal adhesion kinase 1; focal adhesion kinase isoform FAK-Del33; Focal adhesion kinase-related nonkinase; FRNK; p125FAK; pp125 PTK2; pp125FAK; PPP1R71; Protein phosphatase 1 regulatory subunit 71; protein phosphatase 1, regulatory subunit 71; Protein-tyrosine kinase 2; PTK2 protein tyrosine kinase 2
Gene Aliases: FAK; FAK1; PTK2
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support