Invitrogen
This Antibody was verified by Cell treatment to ensure that the antibody binds to the antigen stated. View Details
Interacts with zinc finger protein A20/TNFAIP3 and inhibits TNF-induced NF-kappa-B-dependent gene expression by interfering with an RIP-or TRAF2-mediated transactivation signal. Increases cell surface CD4(T4) antigen expression. Interacts with HIV-1 matrix protein and is packaged into virions and overexpression can inhibit viral replication. May regulate matrix nuclear localization, both nuclear import of PIC (Preintegration complex) and export of GAG polyprotein and viral genomic RNA during virion production.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 2.4.1.11; Glycogen [starch] synthase, muscle; glycogen synthase; glycogen synthase 1 (muscle); glycogen synthase 3, brain
Gene Aliases: GSY; GYS; GYS1; Gys3; MGS
UniProt ID: (Human) P13807, (Rat) A2RRU1, (Mouse) Q9Z1E4
Entrez Gene ID: (Human) 2997, (Rat) 690987, (Mouse) 14936
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support