Invitrogen
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:1,000 | View 2 publications 2 publications |
Product Specifications | |
---|---|
Species Reactivity |
Bovine, Human, Mouse, Non-human primate, Rat |
Published species |
Mouse |
Host/Isotype |
Rabbit / IgG |
Class |
Monoclonal |
Type |
Antibody |
Clone |
J.10.3 |
Immunogen |
Synthetic phosphopeptide corresponding to residues surrounding pSer177 of IKKß |
Conjugate |
Unconjugated |
Form |
Liquid |
Purification |
Affinity chromatography |
Storage buffer |
0.01M HEPES, pH 7.5, with 0.15M NaCl, 100µg/mL BSA, 50% glycerol |
Contains |
<0.02% sodium azide |
Storage conditions |
-20°C |
RRID |
AB_10986824 |
It is not recommended to aliquot this antibody.
IKK alpha/beta is encoded by CHUK (conserved helix-loop-helix ubiquitous kinase) and IKKB (inhibitor of kappa light polypeptide gene enhancer in B-cells) gene located in chromosome 10 and 8 respectively in humans. Upon stimulation by agents that activate the NF-kB pathway such as TNF alpha, the two subunits of IKK complex, IKK alpha and IKK -beta alongside a regulator NEMO (IKK gamma) phosphorylates IkB proteins. This targets the later for ubiquitin mediated degradation and phosphorylation of NF-kB leading to subsequent translocation to the nucleus. Phosphorylation at serine 176 and 180 located in the activation loop causes a confirmation change that is required for the activation of the kinases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Conserved helix-loop-helix ubiquitous kinase; EC 2.7.11.10; I kappa B kinase 1; i kappa-B kinase alpha; I-kappa-B kinase 1; I-kappa-B kinase 2; I-kappa-B kinase alpha; I-kappa-B kinase-alpha; I-kappa-B-kinase beta; IkappaB kinase; IkappaB kinase alpha; IkB kinase alpha subunit; ikBKA; IKK alpha; IKK-A; IKK-a kinase; IKK-alpha; IKK-B; IKK-beta; IKK1; IKK2; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta; inhibitor of kappaB kinase beta; inhibitor of nuclear factor kappa B kinase beta subunit; Inhibitor of nuclear factor kappa-B kinase subunit alpha; Inhibitor of nuclear factor kappa-B kinase subunit beta; NFKBIKA; NFKBIKB; Nuclear factor NF-kappa-B inhibitor kinase alpha; Nuclear factor NF-kappa-B inhibitor kinase beta; Nuclear factor NFkappaB inhibitor kinase alpha; Serine/threonine protein kinase IKBKB; TCF-16; TCF17; Transcription factor 16
Gene Aliases: AI132552; AI256658; AIM-1; CHUK; Chuk1; Fbx24; Fbxo24; IKBKA; IKBKB; IKK-2; IKK-alpha; IKK-beta; IKK1; IKK2; IKK[b]; IKKA; IKKB; IKKbeta; IMD15; NFKBIKA; NFKBIKB; TCF16
UniProt ID: (Bovine) Q95KV1, (Human) O15111, (Bovine) Q95KV0, (Human) O14920, (Mouse) O88351, (Rat) Q9QY78
Entrez Gene ID: (Bovine) 281073, (Human) 1147, (Mouse) 12675, (Rat) 309361, (Bovine) 281854, (Human) 3551, (Mouse) 16150, (Rat) 84351
Molecular Function:
non-receptor serine/threonine protein kinase
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support