Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
Assay-dependent | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
A synthetic phosphopeptide corresponding to residues surrounding Ser376 of human IKK-gamma protein |
Conjugate |
Unconjugated |
Form |
Liquid |
Storage conditions |
-20°C |
IKK gamma is the regulatory subunit of the inhibitor of the kappaB kinase (IKK) complex, which activates NF-kappaB resulting in activation of genes involved in inflammation, immunity, cell survival, and other pathways. Mutations in this gene result in incontinentia pigmenti, hypohidrotic ectodermal dysplasia, and several other types of immunodeficiencies. Multiple transcript variants encoding different isoforms have been found for this gene. A pseudogene highly similar to this locus is located in an adjacent region of the X chromosome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FIP-3; gikk; I-kappa-B kinase subunit gamma; ikB kinase subunit gamma; IkB kinase-associated protein 1; IKK3; IKKAP1; incontinentia pigmenti; Inhibitor of nuclear factor kappa-B kinase subunit gamma; mF; NEMO; NF-kappa-B essential modifier; NF-kappa-B essential modulator; OTTHUMP00000207079; OTTHUMP00000212188
Gene Aliases: FIP3; IKBKG; NEMO
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support