Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MECP2 Antibodies

          Invitrogen

          Phospho-MECP2 (Ser421) Polyclonal Antibody

          5 Published Figures
          4 References
          View all (48) MECP2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-MECP2 (Ser421) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (5)
          Phospho-MECP2 (Ser421) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-MECP2 (Ser421) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-MECP2 (Ser421) Antibody (PA5-35396) in WB

          Western blot of MECP2 in mouse whole brain lysate showing specific immunolabeling of a band at ~55 kDa corresponding to truncated Phospho-MECP2 (Ser421) polyclonal antibody (Product # PA5-35396) in the first lane (-). Phosphospecificity is shown in the second lane (+) where immunolabeling is blocked by pre-adsorption of the phosphopeptide used as the antigen, but not by the corresponding non-phosphopeptide ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-MECP2 (Ser421) Antibody in Western Blot (WB)
          Phospho-MECP2 (Ser421) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-MECP2 (Ser421) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-MECP2 (Ser421) Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Phospho-MECP2 (Ser421) Antibody in Western Blot (WB)
          Phospho-MECP2 (Ser421) Antibody in Western Blot (WB)
          Phospho-MECP2 (Ser421) Polyclonal Antibody

          Product Details

          PA5-35396

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 2 publications 2 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser421 of mouse MeCP2 conjugated to KLH
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          0.01M HEPES, pH 7.5, with 0.1mg/mL BSA, 50% glycerol, 0.15M NaCl

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2552706

          Product Specific Information

          This antibody is predicted to react with human and non-human primate based on 100% sequence homology.

          This antibody contains enough material to conduct 10 mini-Western blots.

          Target Information

          MECP2 belongs to a family of nuclear proteins (including MBD1, MBD2, MBD3 and MBD4) that have a methyl-CpG binding domain. MECP2 is capable of binding specifically to methylated DNA. DNA methylation is the major modification of eukaryotic genomes and plays an essential role in mammalian development. MECP2 can also repress transcription from methylated gene promoters. In contrast to other MBD family members, MECP2 is X-linked and subject to X inactivation. MECP2 is dispensible in stem cells, but is essential for embryonic development. MECP2 gene mutations are the cause of most cases of Rett syndrome, a progressive neurologic developmental disorder and one of the most common causes of mental retardation in females.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: DKFZp686A24160; MeCp-2 protein; Methyl-CpG-binding protein 2

          View more View less

          Gene Aliases: 1500041B07Rik; D630021H01Rik; Mbd5; Mecp2; WBP10

          View more View less

          UniProt ID: (Mouse) Q9Z2D6, (Rat) Q00566

          View more View less

          Entrez Gene ID: (Mouse) 17257, (Rat) 29386

          View more View less

          Function(s)
          four-way junction DNA binding DNA binding chromatin binding transcription factor activity, sequence-specific DNA binding mRNA binding protein binding transcription factor binding methyl-CpG binding double-stranded methylated DNA binding enzyme binding protein domain specific binding chromatin DNA binding siRNA binding histone deacetylase binding poly(A) RNA binding unmethylated CpG binding protein N-terminus binding DNA metabolism protein
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter behavioral fear response response to hypoxia startle response neurological system process involved in regulation of systemic arterial blood pressure regulation of respiratory gaseous exchange by neurological system process inositol metabolic process mitochondrial electron transport, ubiquinol to cytochrome c chromatin silencing regulation of gene expression by genetic imprinting transcription, DNA-templated regulation of transcription, DNA-templated glutamine metabolic process cellular biogenic amine metabolic process chemical synaptic transmission synapse assembly brain development respiratory gaseous exchange learning memory long-term memory protein localization glucocorticoid metabolic process positive regulation of cell proliferation adult locomotory behavior visual learning pathogenesis embryo development post-embryonic development regulation of gene expression dendrite development histone methylation histone acetylation proprioception sensory perception of pain cerebellum development ventricular system development neuron differentiation negative regulation of histone methylation neuron projection development cardiolipin metabolic process multicellular organismal response to stress negative regulation of histone acetylation social behavior regulation of gene expression, epigenetic neuron maturation negative regulation of neuron apoptotic process regulation of DNA methylation negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated phosphatidylcholine metabolic process regulation of synaptic plasticity negative regulation of astrocyte differentiation catecholamine secretion neuromuscular process controlling posture neuromuscular process positive regulation of synapse assembly excitatory postsynaptic potential long-term synaptic potentiation heart development response to ionizing radiation response to lead ion response to organic cyclic compound response to estradiol
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-bxd7r:80/100.66.79.78:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline