Invitrogen
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:1,000 | View 7 publications 7 publications |
Immunocytochemistry (ICC/IF) |
- | View 1 publication 1 publication |
Product Specifications | |
---|---|
Species Reactivity |
Bovine, Chicken, Human, Mouse, Pig, Rat, Xenopus, Zebrafish |
Published species |
Bovine, Human, Mouse |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Synthetic phosphopeptide corresponding to residues surrounding pThr18/Ser19 of human myosin light chain 2 |
Conjugate |
Unconjugated |
Form |
Liquid |
Purification |
Antigen affinity chromatography |
Storage buffer |
0.01M HEPES, pH 7.5, with 0.15M NaCl, 100µg/mL BSA, 50% glycerol |
Contains |
no preservative |
Storage conditions |
-20°C |
RRID |
AB_10980950 |
It is not recommended to aliquot this antibody.
This antibody is not cross-reactive with the cardiac isoform of myosin light chain 2.
Myosin, a structural component of muscle, consists of two heavy chains and four light chains. Myosin light chain may regulate muscle contraction by modulating the ATPase activity of myosin heads. It also binds calcium and is activated by myosin light chain kinase. Two transcript variants encoding different isoforms have been found.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 20 kDa myosin light chain; DTNB; G1; Isoform L20-A; LC20; light polypeptide 9; MGC3505; MLC-2; MLC-2C; myl9l; myosin light chain 2; myosin light chain, regulatory C; myosin regulatory light chain; myosin regulatory light chain 1; Myosin regulatory light chain 2, smooth muscle isoform; Myosin regulatory light chain 2, smooth muscle major isoform; Myosin regulatory light chain 9; Myosin regulatory light chain MRLC1; Myosin regulatory light polypeptide 9; Myosin RLC; myosin, light chain 9, regulatory; myosin, light polypeptide 9, regulatory; smooth muscle major isoform; unnamed protein product
Gene Aliases: AI327049; DTNB; G1; LC20; MLC-2; MLC-2C; MLC2; MLC20; MRLC1; MYL9; Mylc2c; MYRL2; RLC-C; XELAEV_18003093mg; zgc:77916
UniProt ID: (Pig) P29269, (Chicken) P02612, (Human) P24844, (Rat) Q64122, (Mouse) Q3TW15
Entrez Gene ID: (Pig) 100157760, (Chicken) 396215, (Bovine) 505964, (Human) 10398, (Rat) 296313, (Mouse) 98932, (Xenopus) 446851, (Zebrafish) 406493
Molecular Function:
actin or actin-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support