Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
- | View 1 publication 1 publication |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:50-1:100 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse |
Published species |
Mouse |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
A synthetic phosphopeptide derived from human NFAT2 around the phosphorylation site of Ser294 (R-V-SP-V-T) |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
Dulbecco's PBS, pH 7.4, with 150mM NaCl, 50% glycerol |
Contains |
0.02% sodium azide |
Storage conditions |
-20°C |
RRID |
AB_2554902 |
The product of this gene is a component of the nuclear factor of activated T cells DNA-binding transcription complex. This complex consists of at least two components: a preexisting cytosolic component that translocates to the nucleus upon T cell receptor (TCR) stimulation, and an inducible nuclear component. Proteins belonging to this family of transcription factors play a central role in inducible gene transcription during immune response. The product of this gene is an inducible nuclear component. It functions as a major molecular target for the immunosuppressive drugs such as cyclosporin A. Five transcript variants encoding distinct isoforms have been identified for this gene. Different isoforms of this protein may regulate inducible expression of different cytokine genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC138448; NF-ATc; NF-ATc1; NFAT tra; NFAT trans; NFAT transcription complex cytosolic component; nuclear factor of activated T-cells 'c'; Nuclear factor of activated T-cells, cytoplasmic 1; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 1; transcription factor NF-ATc
Gene Aliases: 2210017P03Rik; AI449492; AV076380; NF-ATC; NF-ATc1.2; NFAT2; NFATC; NFATC1; Nfatcb
UniProt ID: (Human) O95644
Entrez Gene ID: (Human) 4772, (Mouse) 18018
Molecular Function:
Rel homology transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support