Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:1,000 | - |
Dot blot (DB) |
1:1,000 | - |
Product Specifications | |
---|---|
Species Reactivity |
Rat |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Tyr1472 of rat GRIN2B conjugated to KLH |
Conjugate |
Unconjugated |
Form |
Liquid |
Purification |
Affinity chromatography |
Storage buffer |
0.01M HEPES, pH 7.5, with 0.15M NaCl, 100µg/mL BSA, 50% glycerol |
Contains |
no preservative |
Storage conditions |
-20°C or -80°C if preferred |
RRID |
AB_325370 |
This antibody is predicted to react with bovine, canine, chicken, human, mouse, non-human primate and zebrafish based on 100% sequence homology.
In Western blot, this antibody detects a ~180 kDa protein representing NMDAR NR2B-subunit protein phosphorylated at Tyr1472. It also labels proteins of ~65k and ~115k. Immunolabeling is completely blocked by blocked by either lamda-Ptase or by the phosphopeptide used as the antigen but not by the corresponding dephosphopeptide.
NMDAR2B is a member of the N-methyl-D-aspartate (NMDA) class of ionotropic glutamate receptors. NMDA receptors (NMDAR) have been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDAR play a role in neuronal development and has been implicated in several disorders of the central nervous system including Alzheimer's Disease, epilepsy and ischemic neuronal cell death. Overexpression of the NR2B subunit of the receptor has been associated with increases in learning and memory while aged, memory impaired animals have deficiencies in NR2B expression. Tyr1472 on NR2B is phosphorylated and this may lead to the increased expression of the NMDAR at the synapse that plays a role in synaptic plasticity. NMDR2B is a heterotrimer comprised of seven enetically encoded and differentially expressed subunits that include NR1, NR2 and NR3. Diseases associated with the dysfunction of NMDAR2B include epileptic encephalopathy, autosomal dominant 6-related mental retardation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GluN2B; Glutamate [NMDA] receptor subunit epsilon-2; glutamate receptor; Glutamate receptor ionotropic, NMDA 2B; glutamate receptor, ionotropic, NMDA2B; GRIN 2B; HGNC:4586; hNR 3; MGC142178; MGC142180; N-methyl D-aspartate receptor subtype 2B; NMDAR2B; NMDE2; NME2; NR2B
Gene Aliases: GluN2B; Grin2b
UniProt ID: (Rat) Q00960
Entrez Gene ID: (Rat) 24410
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support